Skip to Content
Merck
All Photos(1)

Key Documents

SML2254

Sigma-Aldrich

J-113397

≥98% (HPLC)

Synonym(s):

1-[(3R,4R)-1-(Cyclooctylmethyl)-3-(hydroxymethyl)-4-piperidinyl]-3-ethyl-1,3-dihydro-2H-benzimidazol-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$208.00
25 MG
$837.00

$208.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 MG
$208.00
25 MG
$837.00

About This Item

Empirical Formula (Hill Notation):
C24H37N3O2
CAS Number:
Molecular Weight:
399.57
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$208.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +7 to +10°, c = 1 in isopropanol

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

CCN1C2=CC=CC=C2N([C@H]3[C@H](CO)CN(CC4CCCCCCC4)CC3)C1=O

InChI

1S/C24H37N3O2/c1-2-26-22-12-8-9-13-23(22)27(24(26)29)21-14-15-25(17-20(21)18-28)16-19-10-6-4-3-5-7-11-19/h8-9,12-13,19-21,28H,2-7,10-11,14-18H2,1H3/t20-,21+/m0/s1

InChI key

MBGVUMXBUGIIBQ-LEWJYISDSA-N

Biochem/physiol Actions

Antagonist of NOP, the nociceptin/orphanin FQ (N/OFQ) receptor.
J-113397 is an antagonist of NOP, the nociceptin/orphanin FQ (N/OFQ) receptor. J-113397 exhibited an IC50 value of 2.3 nM in Chinese hamster ovary (CHO) cells expressing ORL1 receptor showed with 600-fold or less affinity for μ-, δ- and κ-opioid receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raymond F Genovese et al.
Behavioural pharmacology, 28(7), 521-530 (2017-07-14)
The nociceptin/orphanin FQ peptide (NOP) receptor is believed to have an integral modulatory function in the stress response system. We evaluated the highly selective NOP antagonist J-113397 (7.5 and 20.0 mg/kg), using a predator exposure in which rats were exposed to
Klaus Linz et al.
Anesthesiology, 126(4), 708-715 (2017-03-16)
Cebranopadol is a first-in-class analgesic with agonist activity at classic opioid peptide receptors and the nociceptin/orphanin FQ peptide receptor. The authors compared the antinociceptive and respiratory depressant effects of cebranopadol and the classic opioid fentanyl and used selective antagonists to
S Ozaki et al.
European journal of pharmacology, 387(3), R17-R18 (2000-01-29)
We discovered a potent nociceptin/orphanin FQ receptor (ORL1) receptor antagonist, J-113397 (1-[(3R, 4R)-1-cyclooctylmethyl-3-hydroxymethyl-4-piperidyl]-3-ethyl-1, 3-dihydro-2H-benzimidazol-2-one). J-113397 inhibited [125I][Tyr(14)]nociceptin binding to Chinese hamster ovary (CHO) cells expressing ORL1 receptor in a dose-dependent manner (IC(50); 2. 3 nM), but showed 600-fold or less

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service