Skip to Content
Merck
All Photos(1)

Key Documents

SML2146

Sigma-Aldrich

SR-18292 Maleate

≥98% (HPLC)

Synonym(s):

SR-18292 Maleate, 1-[(1,1-Dimethylethyl)[(4-methylphenyl)methyl]amino]-3-(1H-indol-4-yloxy)-2-propanol maleate, SR 18292, SR 18292 maleate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$237.00
25 MG
$604.00

$237.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$237.00
25 MG
$604.00

About This Item

Empirical Formula (Hill Notation):
C23H30N2O2 · C4H4O4
CAS Number:
Molecular Weight:
482.57
UNSPSC Code:
12352200
NACRES:
NA.77

$237.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C)(C)N(CC(O)COC1=CC=CC2=C1C=CN2)CC3=CC=C(C)C=C3.O=C(O)/C=C\C(O)=O

InChI key

CMTOYDGVFTZNSM-BTJKTKAUSA-N

Application

SR-18292 Maleate has been used as a peroxisome proliferator-activated receptor γ coactivator 1 α (PGC1α) inhibitor to study the protective effects of pioglitazone on demyelinated axons.[1]

Biochem/physiol Actions

SR-18292 is a potent and specific inhibitor of hepatic gluconeogenesis via increased acetylation of PGC-1α and suppression of gluconeogenic gene expression. SR-18292 reduces blood glucose and increases hepatic insulin sensitivity in mouse models of T2D.
inhibitor of hepatic gluconeogenesis via increased acetylation of PGC-1α; anti-diabetic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Licht-Mayer et al.
Acta neuropathologica, 140(2), 143-167 (2020-06-24)
Axonal loss is the key pathological substrate of neurological disability in demyelinating disorders, including multiple sclerosis (MS). However, the consequences of demyelination on neuronal and axonal biology are poorly understood. The abundance of mitochondria in demyelinated axons in MS raises
Kfir Sharabi et al.
Cell, 169(1), 148-160 (2017-03-25)
Type 2 diabetes (T2D) is a worldwide epidemic with a medical need for additional targeted therapies. Suppression of hepatic glucose production (HGP) effectively ameliorates diabetes and can be exploited for its treatment. We hypothesized that targeting PGC-1α acetylation in the liver
Rama Krishna Nimmakayala et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 27(19), 5415-5429 (2021-06-27)
Metabolic reprogramming and cancer stem cells drive the aggressiveness of pancreatic ductal adenocarcinoma (PDAC). However, the metabolic and stemness programs of pancreatic precursor lesions (PPL), considered early PDAC development events, have not been thoroughly explored. Meta-analyses using gene expression profile
Prakash P Praharaj et al.
Autophagy, 20(6), 1359-1382 (2024-03-07)
Mitophagy involves the selective elimination of defective mitochondria during chemotherapeutic stress to maintain mitochondrial homeostasis and sustain cancer growth. Here, we showed that CLU (clusterin) is localized to mitochondria to induce mitophagy controlling mitochondrial damage in oral cancer cells. Moreover

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service