Skip to Content
Merck
All Photos(1)

Key Documents

SML0802

Sigma-Aldrich

ZLN005

≥95% (HPLC)

Synonym(s):

2-(4-tert-Butylphenyl)-1H-benzimidazole, 2-[4-(1,1-Dimethylethyl)phenyl]-1H-benzimidazole

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$98.10
50 MG
$380.00

$98.10


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$98.10
50 MG
$380.00

About This Item

Empirical Formula (Hill Notation):
C17H18N2
CAS Number:
Molecular Weight:
250.34
UNSPSC Code:
12352200
NACRES:
NA.77

$98.10


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

[nH]1c2c(nc1c3ccc(cc3)C(C)(C)C)cccc2

InChI

1S/C17H18N2/c1-17(2,3)13-10-8-12(9-11-13)16-18-14-6-4-5-7-15(14)19-16/h4-11H,1-3H3,(H,18,19)

InChI key

LQUNNCQSFFKSSK-UHFFFAOYSA-N

Biochem/physiol Actions

ZLN005 is a transcriptional regulator of peroxisome proliferator-activated receptor-γ coactivator-1α (PGC-1α). ZLN005 increased PGC-1 and downstream gene transcription in skeletal muscle, while reducing hepatic PGC-1α and gluconeogenesis genes. ZLN005 increased fat oxidation and improved the glucose tolerance, pyruvate tolerance, insulin sensitivity, hyperglycemia and dyslipidemia of diabetic db/db mice.
ZLN005 is a transcriptional regulator of peroxisome proliferator-activated receptor-γ coactivator-1α.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tianyu Hang et al.
International journal of molecular sciences, 24(10) (2023-05-27)
Type-2 diabetes (T2DM) and arterial hypertension (HTN) are major risk factors for heart failure. Importantly, these pathologies could induce synergetic alterations in the heart, and the discovery of key common molecular signaling may suggest new targets for therapy. Intraoperative cardiac

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service