Skip to Content
Merck
All Photos(1)

Key Documents

SML1920

Sigma-Aldrich

ML364

≥98% (HPLC)

Synonym(s):

2-(4-Methylphenylsulfonamido)-N-(4-phenylthiazol-2-yl)-4-(trifluoromethyl)benzamide, 2-[[(4-Methylphenyl)sulfonyl]amino]-N-(4-phenyl-2-thiazolyl)-4-(trifluoromethyl)-benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$112.00
25 MG
$362.00

$112.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$112.00
25 MG
$362.00

About This Item

Empirical Formula (Hill Notation):
C24H18F3N3O3S2
CAS Number:
Molecular Weight:
517.54
UNSPSC Code:
12352200
NACRES:
NA.77

$112.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(c(cc1)C(=O)Nc3[s]cc(n3)c4ccccc4)N[S](=O)(=O)c2ccc(cc2)C

InChI key

QZUGMNXETPARLI-UHFFFAOYSA-N

Biochem/physiol Actions

ML364 is a potent and specific inhibitor of the deubiquitinase USP2 that induces cellular Cyclin D1 degradation in HCT116 colorectal cancer cells, followed by cell cycle arrest and apoptosis. Also, ML364 inhibits homologous recombination (HR)-mediated DNA repair.
Potent and specific inhibitor of the deubiquitinase USP2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mindy I Davis et al.
The Journal of biological chemistry, 291(47), 24628-24640 (2016-09-30)
Deubiquitinases are important components of the protein degradation regulatory network. We report the discovery of ML364, a small molecule inhibitor of the deubiquitinase USP2 and its use to interrogate the biology of USP2 and its putative substrate cyclin D1. ML364

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service