Skip to Content
Merck
All Photos(1)

Key Documents

SML3584

Sigma-Aldrich

AZ1

≥98% (HPLC)

Synonym(s):

2-[[[5-Bromo-2-[[4-fluoro-3-(trifluoromethyl)phenyl]methoxy]phenyl]methyl]amino]-ethanol, AZ 1, AZ-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16BrF4NO2
CAS Number:
Molecular Weight:
422.21
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC=C(COC2=CC=C(C=C2CNCCO)Br)C=C1C(F)(F)F

InChI key

ITHSFXDGKQYOED-UHFFFAOYSA-N

Biochem/physiol Actions

AZ1 is a potent and selective dual USP25 & USP28 deubiquitinase (DUBs) inhibitor (USP25/28 IC50 = 0.62/0.7 nM by Ub-Rh110 assay) that induces HCT116 cellular c-Myc degradation (20-100 µM for 3h or 60 µM for 0.3-3h). AZ1 exhibits antiproliferation activity in cancer cultures (IC50 ≤20 µM; HCT116, HT29 and SW480) and anti-tumor efficacy in an orthotopic model of lung SCC tumours in mice in vivo (125 mg/kg or 375 mg/kg i.p. once every three days, 6 injections total).
Potent and selective dual USP25 & USP28 deubiquitinase (DUBs) inhibitor with anti-cancer efficacy in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maintaining protein stability of ?Np63 via USP28 is required by squamous cancer cells
Cristian Prieto-Garcia, Oliver Hartmann, Michaela Reissland, Fabian Braun, Thomas Fischer
EMBO Molecular Medicine, 12 (2022)
USP28 enables oncogenic transformation of respiratory cells, and its inhibition potentiates molecular therapy targeting mutant EGFR, BRAF and PI3K
Molecular Oncology, 16, 3082-3106 (2022)
Inhibition of USP28 overcomes Cisplatin-resistance of squamous tumors by suppression of the Fanconi anemia pathway
Cell Death and Differentiation, 29, 568-584 (2022)
The deubiquitinase USP25 supports colonic inflammation and bacterial infection and promotes colorectal cancer
Nature cancer, 1, 811-825 (2020)
Deubiquitylase USP25 prevents degradation of BCR-ABL protein and ensures proliferation of Ph-positive leukemia cells
Oncogene, 39, 3867-3878 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service