Skip to Content
Merck
All Photos(1)

Key Documents

SML1911

Sigma-Aldrich

INI-43

≥98% (HPLC)

Synonym(s):

2-Amino-3-(1H-benzimidazol-2-yl)-N,N-dimethyl-1H-pyrrolo[2,3-b]quinoxaline-1-propanamine, 3-(1H-Benzimidazol-2-yl)-1-(3-dimethylaminopropyl)pyrrolo[5,4-b]quinoxalin-2-amine, Inhibitor of Nuclear Import-43

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$148.00
25 MG
$597.00

$148.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$148.00
25 MG
$597.00

About This Item

Empirical Formula (Hill Notation):
C22H23N7
CAS Number:
Molecular Weight:
385.46
UNSPSC Code:
12352200
NACRES:
NA.77

$148.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C)CCCN1C(N)=C(C2=NC3=C(C=CC=C3)N2)C(C1=N4)=NC5=C4C=CC=C5

InChI

1S/C22H23N7/c1-28(2)12-7-13-29-20(23)18(21-25-15-9-4-5-10-16(15)26-21)19-22(29)27-17-11-6-3-8-14(17)24-19/h3-6,8-11H,7,12-13,23H2,1-2H3,(H,25,26)

InChI key

LWPQQQAILAUWTI-UHFFFAOYSA-N

Biochem/physiol Actions

INI-43 is a cell penetrant and potent inhibitor of Kpnb1-mediated nuclear import that cancer cell death via a G2–M cell cycle arrest followed by apoptosis. INI-43 inhibits the nuclear localization of Kpnb1 as well as that of its cargo transcription factors, NFY, AP-1, p65, and NFAT. INI-43 exhibit specific cytotoxicity toward cancer cells. INI-43 inhibits tumor growth in cancer xenograft models.
INI-43 is a cell penetrant and potent inhibitor of Kpnb1-mediated nuclear import.
INI-43 is also known as (3-(1H-benzimidazol-2-yl)-1-(3-dimethylaminopropyl)pyrrolo[5,4-b]quinoxalin-2-amine).[1] It has the ability to prevent the development of dermatologically xenografted esophageal and cervical tumor cells.[1] INI-43 can also decrease activator protein 1 (AP-1) transcriptional activity, induced by phorbol-12-myristate-13-acetate (PMA).[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

KPNB1-mediated nuclear import is required for motility and inflammatory transcription factor activity in cervical cancer cells
Stelma T, et al.
Oncotarget, 8(20), 32833-32833 (2017)
Targeting the nuclear import receptor, Kpn beta as an anti-cancer therapeutic
Watt VD, et al.
Molecular Cancer Therapeutics, molcanther-molcan0052 (2016)
Pauline J van der Watt et al.
Molecular cancer therapeutics, 15(4), 560-573 (2016-02-03)
Karyopherin beta 1 (Kpnβ1) is a nuclear transport receptor that imports cargoes into the nucleus. Recently, elevated Kpnβ1 expression was found in certain cancers and Kpnβ1 silencing with siRNA was shown to induce cancer cell death. This study aimed to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service