Skip to Content
Merck
All Photos(1)

Key Documents

SML0412

Sigma-Aldrich

IM-54

≥98% (HPLC)

Synonym(s):

1-Methyl-3-(1-methyl-1H-indol-3-yl)-4-(pentylamino)-1H-pyrrole-2,5-dione, 2-(1H-Indol-3-yl)-3-pentylamino-maleimide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$247.00

$247.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$247.00

About This Item

Empirical Formula (Hill Notation):
C19H23N3O2
CAS Number:
Molecular Weight:
325.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$247.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

light orange to dark orange

solubility

DMSO: 5 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

CCCCCNC1=C(C(=O)N(C)C1=O)c2cn(C)c3ccccc23

InChI

1S/C19H23N3O2/c1-4-5-8-11-20-17-16(18(23)22(3)19(17)24)14-12-21(2)15-10-7-6-9-13(14)15/h6-7,9-10,12,20H,4-5,8,11H2,1-3H3

InChI key

SGLOMINNEBLJFF-UHFFFAOYSA-N

Biochem/physiol Actions

IM-54 is a cell permeable, potent and selective inhibitor of necrosis.
IM-54 is a cell permeable, potent and selective inhibitor of necrosis. The compound inhibits necrosis induced by oxidative stress. IM-54 does not inhibit apoptosis induced by anticancer drugs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Sun et al.
Free radical biology & medicine, 108, 433-444 (2017-04-18)
Necroptosis is a form of programmed necrosis mediated by signaling complexes with receptor-interacting protein 1 (RIP1) and RIP3 kinases as the main mediators. However, the underlying execution pathways of this phenomenon have yet to be elucidated in detail. In this
Mladen Korbelik et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 19(9), 1145-1151 (2020-08-22)
Our recent investigation uncovered that the acid ceramidase inhibitor LCL521 enhances the direct tumor cell killing effect of photodynamic therapy (PDT) treatment. The present study aimed at elucidating the mechanisms underlying this effect. Exposing mouse squamous cell carcinoma SCCVII cells
Longhao Sun et al.
Autophagy, 13(4), 703-714 (2017-01-26)
Pancreatic ductal adenocarcinoma (PDAC) is the most aggressive and lethal cancer. The role of autophagy in the pathobiology of PDAC is intricate, with opposing functions manifested in different cellular contexts. MIR506 functions as a tumor suppressor in many cancer types
Hexin Shi et al.
Nature communications, 12(1), 1379-1379 (2021-03-04)
Many immune responses depend upon activation of NF-κB, an important transcription factor in the elicitation of a cytokine response. Here we show that N4BP1 inhibits TLR-dependent activation of NF-κB by interacting with the NF-κB signaling essential modulator (NEMO, also known
Mabel Lum et al.
International journal of medical microbiology : IJMM, 304(5-6), 530-541 (2014-04-24)
Shigella infection in epithelial cells induces cell death which is accompanied by mitochondrial dysfunction. In this study the role of the mitochondrial fission protein, Drp1 during Shigella infection in HeLa cells was examined. Significant lactate dehydrogenase (LDH) release was detected

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service