Skip to Content
Merck
All Photos(1)

Key Documents

SML1851

Sigma-Aldrich

RI(dl)-2

≥98% (HPLC)

Synonym(s):

N-Ethyl-4-(pyrrolo[1,2-a]quinoxalin-4-yl)aniline

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$112.00
25 MG
$452.00

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$112.00
25 MG
$452.00

About This Item

Empirical Formula (Hill Notation):
C19H17N3 · xC2HF3O2
CAS Number:
Molecular Weight:
287.36 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, light orange to dark red

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCNC1=CC=C(C2=NC3=C(N4C2=CC=C4)C=CC=C3)C=C1.FC(F)(C(O)=O)F

InChI

1S/C19H17N3.C2HF3O2/c1-2-20-15-11-9-14(10-12-15)19-18-8-5-13-22(18)17-7-4-3-6-16(17)21-19;3-2(4,5)1(6)7/h3-13,20H,2H2,1H3;(H,6,7)

InChI key

NIJZNZBFNNPMPH-UHFFFAOYSA-N

Biochem/physiol Actions

RI(dl)-2 is a cell-permeable pyrroloquinoxaline derivative that selectively inhibits Rad51-mediated D-loop formation (IC50 = 11.1 μM) without affecting its ssDNA-binding activity (IC50 >100 μM). RI(dl)-2 is shown to inhibit DSBs-induced cellular homologous recombination (HR) activity (IC50 = 3.0 μM by HEK293-based DR-GFP reporter assay) by stabilizing nucleoprotein filaments in a nonfunctional state without affecting DNA damage-induced RAD51 nuclear loci formation or stimulating single-strand annealing (SSA) activity.
RI(dl)-2 is a cell-permeable pyrroloquinoxaline derivative that selectively inhibits Rad51-mediated D-loop formation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service