Skip to Content
Merck
All Photos(1)

Key Documents

SML1817

Sigma-Aldrich

BCI-121

≥98% (HPLC)

Synonym(s):

4-(Aminocarbonyl)-N-(4-bromophenyl)-1-piperidineacetamide, BCI121

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$122.00
25 MG
$496.00

$122.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 MG
$122.00
25 MG
$496.00

About This Item

Empirical Formula (Hill Notation):
C14H18BrN3O2
CAS Number:
Molecular Weight:
340.22
UNSPSC Code:
12352200
NACRES:
NA.77

$122.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

Brc1ccc(cc1)NC(=O)CN2CCC(CC2)C(=O)N

InChI

1S/C14H18BrN3O2/c15-11-1-3-12(4-2-11)17-13(19)9-18-7-5-10(6-8-18)14(16)20/h1-4,10H,5-9H2,(H2,16,20)(H,17,19)

InChI key

KSUYPIXCRPCPGF-UHFFFAOYSA-N

Biochem/physiol Actions

BCI-121 is a substrate-competitive SMYD3 inhibitor that reduces nuclear histone H3 lys4 di- and tri-methylation level (by 50%/H3K4me2 and 40%H3K4me3 in HT29 cells; 100 μM BCI-121 for 48 h), downregulates known SMYD3 target genes transcription, and selectively affects SMYD3-dependent proliferation of cancer cultures (46%/HT29 and 54%/HCT116 proliferation reduction; 100 μM BCI-121 for 72 h) with little antiproliferation efficacy toward low SMYD3-expressing cancer cells. BCI-121 targets SMYD3 via direct affinity interaction (kon 357.7/M/s; koff 4.23×10-3/s; KD=koff/kon = 11.8 μM) and effectively competes against histone for SMYD3 binding (%inhibition/[histone H4 peptide]:[BCI-121] ratio = 36.5%/1:1 and 51.0%/1:2.5).
BCI-121 is a substrate-competitive SMYD3 inhibitor.
BCI121 is capable of reducing the mesenchymal signature of MDA-MB-231 cells. It can also decrease their ability to invade in vitro and in vivo.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryuji Hamamoto et al.
Cancer science, 107(4), 377-384 (2016-01-12)
Protein methylation is one of the important post-translational modifications. Although its biological and physiological functions were unknown for a long time, we and others have characterized a number of protein methyltransferases, which have unveiled the critical functions of protein methylation
SMYD3 promotes the epithelial-mesenchymal transition in breast cancer
Fenizia C, et al.
Nucleic Acids Research, 47(3), 1278-1293 (2018)
Tianjiao Lyu et al.
International journal of cancer, 146(6), 1553-1567 (2019-09-11)
Detachment of cancer cells from the primary tumor and formation of spheroids in ascites is required for implantation metastasis in epithelial ovarian cancer (EOC), but the underlying mechanism of this process has not been thoroughly elucidated. To mimic this process
Alessia Peserico et al.
Journal of cellular physiology, 230(10), 2447-2460 (2015-03-03)
SMYD3 is a histone lysine methyltransferase that plays an important role in transcriptional activation as a member of an RNA polymerase complex, and its oncogenic role has been described in different cancer types. We studied the expression and activity of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service