Skip to Content
Merck
All Photos(1)

Key Documents

SML1792

Sigma-Aldrich

Oxazole yellow

≥98% (HPLC)

Synonym(s):

4-[(3-Methyl-2(3H)-benzoxazolylidene)methyl]-1-[3-(trimethylammonio)propyl]-quinolinium diiodide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$328.00

$328.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 MG
$328.00

About This Item

Empirical Formula (Hill Notation):
C24H29I2N3O
CAS Number:
Molecular Weight:
629.32
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$328.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

yellow to orange-brown

solubility

H2O: 5, clear (warmed)

storage temp.

2-8°C

SMILES string

CN1C2=CC=CC=C2OC1=CC3=CC=[N+](CCC[N+](C)(C)C)C4=C3C=CC=C4

InChI

1S/C24H29N3O.2HI/c1-25-22-12-7-8-13-23(22)28-24(25)18-19-14-16-26(15-9-17-27(2,3)4)21-11-6-5-10-20(19)21;;/h5-8,10-14,16,18H,9,15,17H2,1-4H3;2*1H/q+2;;/p-2

InChI key

ULHRKLSNHXXJLO-UHFFFAOYSA-L

Biochem/physiol Actions

Dye to detect apoptotic cells
Oxazole yellow is a fluorescent cyanine dye that binds to DNA and is used to detect apoptotic cells. Oxazole yellow does not enter live cells. However, during apoptosis the cytoplasmic membrane becomes slightly permeable, allowing entry of the dye. Oxazole yellow is often used along with propidium iodide (catalog no. P4170), a DNA dead cell stain that does not enter either live or apoptotic cells. The λEx/λEm of Oxazole yellow is 491/509.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas S Biebricher et al.
Nature communications, 6, 7304-7304 (2015-06-19)
DNA intercalators are widely used as fluorescent probes to visualize DNA and DNA transactions in vivo and in vitro. It is well known that they perturb DNA structure and stability, which can in turn influence DNA-processing by proteins. Here we
Sho Fujisawa et al.
Cytotechnology, 66(2), 259-273 (2013-09-26)
Radiofrequency (RF) ablation (RFA) is a minimally invasive treatment for colorectal-cancer liver metastases (CLM) in selected nonsurgical patients. Unlike surgical resection, RFA is not followed by routine pathological examination of the target tumor and the surrounding liver tissue. The aim

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service