Skip to Content
Merck
All Photos(1)

Key Documents

SML1744

Sigma-Aldrich

A-971432

≥95% (HPLC)

Synonym(s):

1-(4-(3,4-Dichlorobenzyloxy)benzyl)azetidine-3-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$143.00
25 MG
$583.00

$143.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 MG
$143.00
25 MG
$583.00

About This Item

Empirical Formula (Hill Notation):
C18H17Cl2NO3
CAS Number:
Molecular Weight:
366.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$143.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 3 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

ClC1=CC(COC2=CC=C(CN3CC(C(O)=O)C3)C=C2)=CC=C1Cl

InChI

1S/C18H17Cl2NO3/c19-16-6-3-13(7-17(16)20)11-24-15-4-1-12(2-5-15)8-21-9-14(10-21)18(22)23/h1-7,14H,8-11H2,(H,22,23)

InChI key

WAAWETUDFSIYSD-UHFFFAOYSA-N

Biochem/physiol Actions

A-971432 is an orally bioavailable, non-clastogenic, azetidinecarboxylate compound that acts as a highly potent and selective sphingosine-1-phosphate receptor 5 agonist (IC50 = 6 nM/S1P5, 362 nM/S1P1 and >10 μM/S1P3 in a radio-ligand binding assay) without affect the activity of 129 protein kinases (IC50 >10 μM). A-971432 is shown to enhance blood-brain barrier integrity and reverse lipid accumulation and age-related cognitive decline in mice in vivo with good pharmacokinetics (t1/2  = 5.7 h; Cmax = 2,500 ng/ml; AUC = 35,000 ng.h/ml at 10 mg/kg, p.o. in CD1 mice). Likewise, oral gavage showed good plasma exposure and no sign of lymphopenia in rats (10, 30, and 100 mg/kg.
A-971432 is an orally bioavailable, non-clastogenic, azetidinecarboxylate compound that acts as a highly potent and selective sphingosine-1-phosphate receptor 5 agonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service