Skip to Content
Merck
All Photos(1)

Key Documents

SML1740

Sigma-Aldrich

FIN56

≥98% (HPLC)

Synonym(s):

N2,N7-Dicyclohexyl-9-(hydroxyimino)-9H-fluorene-2,7-disulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$143.00
25 MG
$583.00

$143.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 MG
$143.00
25 MG
$583.00

About This Item

Empirical Formula (Hill Notation):
C25H31N3O5S2
CAS Number:
Molecular Weight:
517.66
UNSPSC Code:
12352200
NACRES:
NA.77

$143.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

[S](=O)(=O)(NC5CCCCC5)c1cc2c(cc1)c3c(cc(cc3)[S](=O)(=O)NC4CCCCC4)C2N=O

InChI

1S/C25H31N3O5S2/c29-26-25-23-15-19(34(30,31)27-17-7-3-1-4-8-17)11-13-21(23)22-14-12-20(16-24(22)25)35(32,33)28-18-9-5-2-6-10-18/h11-18,25,27-28H,1-10H2

InChI key

OYLSCKVHQYAZEE-UHFFFAOYSA-N

Biochem/physiol Actions

FIN56 is a specific inducer of ferroptosis, a non-apoptotic form of cell death characterized by iron-dependent accumulation of lipid peroxidation products and lethal reactive oxygen species (ROS). FIN56 has an EC50 value of 240 nM, and is believed to act though two separate pathways. The first requires the enzymatic activity of acetyl-CoA carboxylase (ACC),and results in degradation of glutathione peroxidase 4 (GPX4), which acts as a negative regulator of ferroptosis by reducing lipid hydroperoxides. The second pathway involves FIN56 binding to and activing squalene synthase, which leads to coenzyme Q10 depletion.
FIN56 is a specific inducer of ferroptosis.
Ferroptosis inducer 56 (FIN56) is a member of the class 3 ferroptosis inducers.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Unsolved mysteries: How does lipid peroxidation cause ferroptosis?.
Feng H and Stockwell BR
PLoS Biology, 16(5), e2006203-e2006203 (2018)
Xingzhe Ma et al.
Cell metabolism, 33(5), 1001-1012 (2021-03-11)
Understanding the mechanisms underlying how T cells become dysfunctional in a tumor microenvironment (TME) will greatly benefit cancer immunotherapy. We found that increased CD36 expression in tumor-infiltrating CD8+ T cells, which was induced by TME cholesterol, was associated with tumor progression and
Po-Han Chen et al.
Cell death and differentiation, 27(3), 1008-1022 (2019-07-20)
Ferroptosis is a specialized iron-dependent cell death that is associated with lethal lipid peroxidation. Modulation of ferroptosis may have therapeutic potential since it has been implicated in various human diseases as well as potential antitumor activities. However, much remains unknown
Ye Zhu et al.
Nature cell biology, 26(4), 542-551 (2024-03-08)
β-Propeller protein-associated neurodegeneration (BPAN) is a rare X-linked dominant disease, one of several conditions that manifest with neurodegeneration and brain iron accumulation. Mutations in the WD repeat domain 45 (WDR45) gene encoding WIPI4 lead to loss of function in BPAN

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service