Skip to Content
Merck
All Photos(1)

Key Documents

SML1736

Sigma-Aldrich

Nelarabine

≥98% (HPLC)

Synonym(s):

2-amino-9-β-D-arabinofuranosyl-6-methoxy-9H-purine, 506U, 506U78, 9-β-D-Arabinofuranosyl-6-methoxy-9H-purin-2-amine, Arranon, Atriance, GW-506U78

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$143.00
50 MG
$583.00

$143.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025



Select a Size

Change View
10 MG
$143.00
50 MG
$583.00

About This Item

Empirical Formula (Hill Notation):
C11H15N5O5
CAS Number:
Molecular Weight:
297.27
UNSPSC Code:
12352200
NACRES:
NA.77

$143.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](O[C@H](CO)[C@H]1O)N2C=NC3=C2N=C(N)N=C3OC

InChI

1S/C11H15N5O5/c1-20-9-5-8(14-11(12)15-9)16(3-13-5)10-7(19)6(18)4(2-17)21-10/h3-4,6-7,10,17-19H,2H2,1H3,(H2,12,14,15)/t4-,6-,7+,10?/m1/s1

InChI key

IXOXBSCIXZEQEQ-KBNQYOMWSA-N

Biochem/physiol Actions

Antineoplastic purine nucleoside analog used in T-cell acute lymphoblastic leukemia; pro-drug of Ara-G.
Nelarabine is an antineoplastic purine nucleoside analog used in T-cell acute lymphoblastic leukemia. It is a pro-drug of Ara-G, which is converted by cellular kinases to the active 5′-triphosphate, Ara-GTP. Incorporation of Ara-GTP into DNA leads to inhibition of DNA synthesis and apoptosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Nelarabine, cyclosphosphamide and etoposide for adults with relapsed T-cell acute lymphoblastic leukaemia and lymphoma.
Marlise R Luskin et al.
British journal of haematology, 174(2), 332-334 (2015-09-26)
Larry W Buie et al.
Clinical therapeutics, 29(9), 1887-1899 (2007-11-24)
Nelarabine was approved by the US Food and Drug Administration (FDA) in October 2005 for the treatment of T-cell acute lymphoblastic leukemia (T-ALL) and T-cell lymphoblastic lymphoma (T-LBL) that has not responded to or has relapsed after treatment with at
Stuart S Winter et al.
Pediatric blood & cancer, 62(7), 1176-1183 (2015-03-11)
Nelarabine has shown impressive single agent clinical activity in T-cell acute lymphoblastic leukemia (T-ALL), but has been associated with significant neurotoxicities in heavily pre-treated patients. We showed previously that it was safe to add nelarabine to a BFM-86 chemotherapy backbone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service