Skip to Content
Merck
All Photos(1)

Key Documents

SML1629

Sigma-Aldrich

Ezetimibe

≥98% (HPLC)

Synonym(s):

(3R,4S)-1-(4-Fluorophenyl)-3-[(S)-3-(4-fluorophenyl)-3-hydroxypropyl]-4-(4-hydroxyphenyl)azetidin-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$164.00

$164.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$164.00

About This Item

Empirical Formula (Hill Notation):
C24H21F2NO3
CAS Number:
Molecular Weight:
409.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$164.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMF: soluble ≥15 mg/mL
DMSO: soluble ≥15 mg/mL
ethanol: soluble ≥15 mg/mL
methanol: soluble ≥15 mg/mL

storage temp.

2-8°C

SMILES string

FC(C=C1)=CC=C1N([C@H](C2=CC=C(O)C=C2)[C@H]3CC[C@H](O)C4=CC=C(F)C=C4)C3=O

InChI

1S/C24H21F2NO3/c25-17-5-1-15(2-6-17)22(29)14-13-21-23(16-3-11-20(28)12-4-16)27(24(21)30)19-9-7-18(26)8-10-19/h1-12,21-23,28-29H,13-14H2/t21-,22+,23-/m1/s1

InChI key

OLNTVTPDXPETLC-XPWALMASSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ezetimibe inhibits intestinal cholesterol absorption by preventing cholesterol uptake by Niemann-Pick C1 Like 1 (NPC1L1) protein that was found critical for the intestinal absorption of cholesterol. Ezetimibe selectvely blocks cholesterol absorption without inhibiting pancreatic lipolytic enzyme activities in the intestinal lumen.
Ezetimibe is a non statin drug that reduces intestinal cholesterol absorption. In addition, it also has an ability to reduce the risk of cardiovascular events in patients who had had an acute coronary syndrome and whose low-density lipoprotein (LDL) cholesterol values were within guideline recommendations.[1]

Reconstitution

For solubility in aqueous buffer, Ezetimibe should be dissolved in ethanol followed by dilution with the buffer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ezetimibe added to statin therapy after acute coronary syndromes.
Cannon C P, et al.
The New England Journal of Medicine, 372(25), 2387-2397 (2015)
Wang, D.Q.
Annual Review of Physiology, 69, 221-248 (2011)
Garcia-Calvo, et al.
Proceedings of the National Academy of Sciences of the USA, 102, 8132-8137 (2005)
Wojciech G Garbacz et al.
Molecular pharmacology, 94(4), 1145-1154 (2018-07-27)
Cholesterol is essential for numerous biologic functions and processes, but an excess of intracellular cholesterol can be toxic. Intestinal cholesterol absorption is a major determinant of plasma cholesterol level. The liver X receptor (LXR) is a nuclear receptor known for
Jing Yu et al.
Oxidative medicine and cellular longevity, 2020, 4717258-4717258 (2020-01-31)
Oxidative stress and neuroinflammation play essential roles in ischemic stroke-induced brain injury. Previous studies have reported that Ezetimibe (Eze) exerts antioxidative stress and anti-inflammatory properties in hepatocytes. In the present study, we investigated the effects of Eze on oxidative stress

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service