Skip to Content
Merck
All Photos(1)

Key Documents

SML1617

Sigma-Aldrich

GSK2033

≥98% (HPLC)

Synonym(s):

2,4,6-Trimethyl-N-[[3′-(methylsulfonyl)[1,1′-biphenyl]-4-yl]methyl]-N-[[5-(trifluoromethyl)-2-furanyl]methyl]benzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$204.00
25 MG
$822.00

$204.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$204.00
25 MG
$822.00

About This Item

Empirical Formula (Hill Notation):
C29H28F3NO5S2
CAS Number:
Molecular Weight:
591.66
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$204.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 3 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CS(C1=CC=CC(C2=CC=C(CN(S(C3=C(C)C=C(C)C=C3C)(=O)=O)CC4=CC=C(C(F)(F)F)O4)C=C2)=C1)(=O)=O

InChI

1S/C29H28F3NO5S2/c1-19-14-20(2)28(21(3)15-19)40(36,37)33(18-25-12-13-27(38-25)29(30,31)32)17-22-8-10-23(11-9-22)24-6-5-7-26(16-24)39(4,34)35/h5-16H,17-18H2,1-4H3

InChI key

PSOXOVKYGWBTPB-UHFFFAOYSA-N

Biochem/physiol Actions

GSK2033 has the ability to repress gluconeogenic gene expression stimulated by GC (glucocorticoids), without altering immune-responsive GR (GC receptor) target genes.[1] It helps to target several nuclear receptors that can alter hepatic gene expression.[2]
GSK2033 is an antagonist of the nuclear receptor liver-X-receptor (LXR) with pIC50 values of 7.0 for LXRα and 7.4 for LXRβ. Treatment of murine CD41 T cells with GSK2033 enhanced cellular proliferation and Th1/Th2/Th17 differentiation. In another study, the conjugated linoleic acid (CLA)-induced expression of efflux protein ATP-binding-cassette transporter A1 (ABCA1) was completely abolished by preincubation with GSK2033, showing that CLA mediated regulation of ABCA-1 expression is LXR dependent.
LXR antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separating the Anti-Inflammatory and Diabetogenic Effects of Glucocorticoids Through LXRβ Antagonism.
Patel R, et al.
Endocrinology, 158(4), 1034-1047 (2017)
Promiscuous activity of the LXR antagonist GSK2033 in a mouse model of fatty liver disease.
Griffett K and Burris TP
Biochemical and Biophysical Research Communications, 479(3), 424-428 (2016)
Lili Chen et al.
Journal of neurochemistry, 154(2), 205-217 (2019-10-12)
Cerebral ischemia (CI) leads to cognitive dysfunction due to the loss of hippocampal neurons. Liver X receptors (LXRs), including the LXRα and LXRβ isoforms, are critical for neurogenesis, synaptic plasticity, neurodegeneration, and cholesterol metabolism. However, the potential role of LXRs
Louise Ménégaut et al.
Cell reports, 31(7), 107665-107665 (2020-05-21)
Low-grade inflammation is constitutive of atherosclerosis, and anti-inflammatory therapy inhibiting interleukin-1β (IL-1β) reduces the rate of cardiovascular events. While cholesterol accumulation in atheroma plaque and macrophages is a major driver of the inflammatory process, the role of the LXR cholesterol

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service