Skip to Content
Merck
All Photos(1)

Key Documents

SML1510

Sigma-Aldrich

SR9238

≥98% (HPLC)

Synonym(s):

Ethyl 5-((2,4,6-trimethyl-N-((3′-(methylsulfonyl)-[1,1′-biphenyl]-4-yl)methyl)phenylsulfonamido)methyl)furan-2-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$135.00
25 MG
$547.00

$135.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$135.00
25 MG
$547.00

About This Item

Empirical Formula (Hill Notation):
C31H33NO7S2
CAS Number:
Molecular Weight:
595.73
UNSPSC Code:
12352200
NACRES:
NA.77

$135.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N(Cc4[o]c(cc4)C(=O)OCC)Cc2ccc(cc2)c3cc(ccc3)[S](=O)(=O)C)c1c(cc(cc1C)C)C

InChI

1S/C31H33NO7S2/c1-6-38-31(33)29-15-14-27(39-29)20-32(41(36,37)30-22(3)16-21(2)17-23(30)4)19-24-10-12-25(13-11-24)26-8-7-9-28(18-26)40(5,34)35/h7-18H,6,19-20H2,1-5H3

InChI key

HDZWHJYZJWLTAG-UHFFFAOYSA-N

Biochem/physiol Actions

SR9238 is liver selective LXR inverse agonist with high potency for both LXRα and LXRβ with an IC50 of 214 nM for LXRα and 43 nM for LXRβ. Metabolic syndrome is often accompanied by liver problems, such as fatty liver (nonalcoholic hepatosteatosis), associated with increased lipogenesis. Liver X receptors α and β (LXRα and LXRβ) are known to induce lipogenesis by increasing transcription of lipogenic enzyme genes, so it was hoped LXR antagonists might be of use in fatty liver. In a mouse model of nonalcoholic hepatosteatosis, SR9238 reduced the expression of lipogenic genes, and suppressed hepatic lipogenesis, inflammation and hepatic lipid accumulation. It was also effective in reduction of hepatic fibrosis in another study. Additionally in diet induced obese mice, SR9238 suppressed plasma cholesterol levels.
SR9238 is liver selective LXR inverse agonist with high potency for both LXRα and LXRβ.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grace Ampem et al.
Cell and tissue research, 378(1), 81-96 (2019-04-24)
Self-renewal of macrophages is important for the healthy development and replenishment of tissue-resident macrophage pools. How this mechanism is controlled by endocrine signals is still largely unexplored. Here, we show that the endocrine disruptor bisphenol A (BPA) increases macrophage self-renewal.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service