Skip to Content
Merck
All Photos(1)

Key Documents

SML2655

Sigma-Aldrich

Reparixin

≥98% (HPLC)

Synonym(s):

αR-methyl-4-(2-methylpropyl)-N-(methylsulfonyl)-benzeneacetamide, (R)-(-)-N-2-[(4-Isobutylphenyl)propionyl]methanesulfonamide, DF 1681Y, Repertaxin

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$176.00
25 MG
$712.00

$176.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$176.00
25 MG
$712.00

About This Item

Empirical Formula (Hill Notation):
C14H21NO3S
CAS Number:
Molecular Weight:
283.39
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$176.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -80 to -90°, c = 1.0 in ethanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[S](=O)(=O)(NC(=O)C(C)c1ccc(cc1)CC(C)C)C

InChI

1S/C14H21NO3S/c1-10(2)9-12-5-7-13(8-6-12)11(3)14(16)15-19(4,17)18/h5-8,10-11H,9H2,1-4H3,(H,15,16)

InChI key

KQDRVXQXKZXMHP-UHFFFAOYSA-N

Biochem/physiol Actions

Noncompetitive allosteric inhibitor of the inflammatory CXCL8 (IL-8) chemokine receptors CXCR1 and CXCR2
Reparixin is a noncompetitive allosteric inhibitor of the inflammatory CXCL8 (IL-8) chemokine receptors CXCR1 and CXCR2 with IC50 values of 1 nM for CXCR1 and 100 nM for CXCR2. It is believed to prevent receptor signaling by locking CXCR1/R2 in an inactive conformation. Reparixin has been studied to prevent ischaemia-reperfusion injury and inflammation and is clinical trials for triple negative breast cancer and to prevent injury occurring after pancreatic islet transplantation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N C Bergenhem et al.
Biochemical genetics, 30(5-6), 279-287 (1992-06-01)
Two, electrophoretically distinct, forms of carbonic anhydrase I (CA Ia and CA Ib) are found at high polymorphic frequencies in red cells of natural populations of pigtail macaques, Macaca nemestrina, from southeast Asia. By use of the polymerase chain reaction
Anne F Schott et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 23(18), 5358-5365 (2017-05-26)
Purpose: Chemokine receptor 1 (CXCR1) is recognized as an actionable receptor selectively expressed by breast cancer stem cells (BCSCs). Reparixin is an investigational allosteric inhibitor of chemokine receptors 1 and 2 (CXCR1/2), and demonstrates activity against BCSCs in human breast
P Opfermann et al.
Clinical and experimental immunology, 180(1), 131-142 (2014-11-18)
Reparixin, a CXCR 1/2 antagonist, has been shown to mitigate ischaemia-reperfusion injury (IRI) in various organ systems in animals, but data in humans are scarce. The aim of this double-blinded, placebo-controlled pilot study was to evaluate the safety and efficacy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service