Skip to Content
Merck
All Photos(1)

Key Documents

SML1380

Sigma-Aldrich

Sacubitril calcium salt

≥98% (HPLC)

Synonym(s):

(2R,4S)-4-[(3-Carboxy-1-oxopropyl)amino]-4-[(p-phenylphenyl)methyl]-2-methylbutanoic acid ethyl ester, 4-[[(2S,4R)-5-Ethoxy-4-methyl-5-oxo-1-(4-phenylphenyl)pentan-2-yl]amino]-4-oxobutanoic acid calcium salt, 4-{[(2S,4R)-1-(4-Biphenylyl)-5-ethoxy-4-methyl-5-oxo-2-pentanyl]amino}-4-oxobutanoic acid; (2R,4S)-5-(Biphenyl-4-yl)-4-[(3-carboxypropionyl)amino]-2-methylpentanoic acid ethyl ester, AHU-377 calcium salt, AHU377 calcium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$144.00
25 MG
$564.00

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$144.00
25 MG
$564.00

About This Item

Empirical Formula (Hill Notation):
C24H28NO5 · 0.5Ca
CAS Number:
Molecular Weight:
430.52
UNSPSC Code:
12352200
NACRES:
NA.77

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

[Ca+2].N([C@@H](C[C@@H](C)C(=O)OCC)Cc3ccc(cc3)c4ccccc4)C(=O)CCC(=O)[O-].N([C@@H](C[C@@H](C)C(=O)OCC)Cc1ccc(cc1)c2ccccc2)C(=O)CCC(=O)[O-]

InChI key

DDLCKLBRBPYKQS-OXXXZDCLSA-L

Biochem/physiol Actions

Sacubitril (AHU377) is a prodrug inhibitor of the membrane metallo-endopeptidase neprilysin.
Sacubitril (AHU377) is a prodrug inhibitor of the membrane metallo-endopeptidase neprilysin. Sacubitril forms half of the heart failure combination drug LCZ696, along with the angiotensin-receptor blocker Valsartan. Sacubitril (AHU377) is cojnverted by esterases into the active inhibitor LBQ657, which prevents neprilysin′s degradation of atrial and brain natriuretic peptide, two blood pressure lowering peptides. The AHU377 valsartan combination drug LCZ696 showed a reduction in the risk of cardiovascular death of 20% compared to the standard ACE inhibitor enalapril, and also a 21% drop in the risk of hospitalization.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. what is the meaning for content of water 2.0 mol? I need it in %, thanks

    1 answer
    1. You would use the ratio of anhydrous molecular weight/hydrated molecular weight. You can then take that value and subtract it from 1. For 2 moles water that would be 430.52/466.52=~0.9228 1-.9228 =0.0772 or 7.72% water.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service