Skip to Content
Merck
All Photos(1)

Key Documents

SML1354

Sigma-Aldrich

Tonabersat

≥98% (HPLC)

Synonym(s):

(3S-cis)-N-(6-Acetyl-3,4-dihydro-3-hydroxy-2,2-dimethyl-2H-1-benzopyran-4-yl)-3-chloro-4-fluoro-benzamide, N-[(3S,4S)-6-Acetyl-3,4-dihydro-3-hydroxy-2,2-dimethyl-2H-1-benzopyran-4-yl]-3-chloro-4-fluoro-benzamide, SB 220453, SB-220453

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$238.00
25 MG
$906.00

$238.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$238.00
25 MG
$906.00

About This Item

Empirical Formula (Hill Notation):
C20H19ClFNO4
CAS Number:
Molecular Weight:
391.82
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$238.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -100 to -110°, c = 1 in methanol

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(C1=CC=C2C([C@H](NC(C3=CC=C(F)C(Cl)=C3)=O)[C@H](O)C(C)(C)O2)=C1)=O

InChI

1S/C20H19ClFNO4/c1-10(24)11-5-7-16-13(8-11)17(18(25)20(2,3)27-16)23-19(26)12-4-6-15(22)14(21)9-12/h4-9,17-18,25H,1-3H3,(H,23,26)/t17-,18-/m0/s1

InChI key

XLIIRNOPGJTBJD-ROUUACIJSA-N

Biochem/physiol Actions

Tonabersat (SB-220453) exhibits anticonvulsant property. It is a member of the family of novel benzoylamino-benzopyran compounds.[1] Tonabersat is used as a therapeutic for trigeminal nerve-induced neurovascular reflexes.[1] In addition, it acts as a putative migraine prophylactic agent containing an unique stereospecific binding site in the brain.[2]
Tonabersat is a potent inhibitor of neuronal-glial gap junctions in trigeminal ganglion that inhibits cortical spreading depression (CSD) and neurogenic inflammation in animal models of migraine.
Tonabersat is a potent inhibitor of neuronal-glial gap junctions.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Randomized, double-blind, placebo-controlled, proof-of-concept study of the cortical spreading depression inhibiting agent tonabersat in migraine prophylaxis.
Goadsby P J, et al.
Cephalalgia, 29(7), 742-750 (2009)
Tonabersat (SB?220453) a novel benzopyran with anticonvulsant properties attenuates trigeminal nerve?induced neurovascular reflexes.
Parsons A A, et al.
British Journal of Pharmacology, 132(7), 1549-1557 (2001)
Esther N Arwert et al.
Nature cell biology, 22(7), 758-766 (2020-06-03)
Cancer-associated fibroblasts (CAFs) perform diverse roles and can modulate therapy responses1. The inflammatory environment within tumours also influences responses to many therapies, including the efficacy of oncolytic viruses2; however, the role of CAFs in this context remains unclear. Furthermore, little

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service