Skip to Content
Merck
All Photos(1)

Key Documents

SML1321

Sigma-Aldrich

ETP-46464

≥98% (HPLC)

Synonym(s):

2-Methyl-2-[4-(2-oxo-9-quinolin-3-yl-4H-[1,3]oxazino[5,4-c]quinolin-1-yl)phenyl]propanenitrile, 4-[4-(1-Isocyano-1-methyl-ethyl)-phenyl]-6-quinolin-3-yl-1,4-dihydro-2-oxa-4,9-diaza-phenanthren-3-one, ATRi

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$156.00

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$156.00

About This Item

Empirical Formula (Hill Notation):
C30H22N4O2
CAS Number:
Molecular Weight:
470.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

CC(C)(C#N)C(C=C1)=CC=C1N(C2=C(CO3)C=NC(C2=C4)=CC=C4C5=CC6=C(C=CC=C6)N=C5)C3=O

InChI

1S/C30H22N4O2/c1-30(2,18-31)23-8-10-24(11-9-23)34-28-22(17-36-29(34)35)16-33-27-12-7-19(14-25(27)28)21-13-20-5-3-4-6-26(20)32-15-21/h3-16H,17H2,1-2H3

InChI key

DPLMXAYKJZOTKO-UHFFFAOYSA-N

Biochem/physiol Actions

ETP-46464 is a potent and selective inhibitor of the DNA damage response kinase Ataxia telangiectasia-mutated (ATM) - and Rad3-related (ATR) with an IC50 of 25 nM. ETP-46464 shows moderate activity against PI3K and is a potent inhibitor of mTor, but is selective for ATR versus ATM and DNA -dependent protein kinase (DNA -PKcs) even at high doses. ETP-46464 inhibited phosphorylation of Chk1, the downstream kinase of ATR, but did not affect ionizing radiation–induced γH2AX formation, which is jointly controlled by ATM and DNA-PKc.
ETP-46464 is a potent and selective inhibitor of the DNA damage response kinase Ataxia telangiectasia-mutated (ATM) - and Rad3-related (ATR).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coline Arnould et al.
Nature, 590(7847), 660-665 (2021-02-19)
The repair of DNA double-strand breaks (DSBs) is essential for safeguarding genome integrity. When a DSB forms, the PI3K-related ATM kinase rapidly triggers the establishment of megabase-sized, chromatin domains decorated with phosphorylated histone H2AX (γH2AX), which act as seeds for
Justin L Sparks et al.
Cell, 176(1-2), 167-181 (2019-01-01)
Covalent DNA-protein cross-links (DPCs) impede replication fork progression and threaten genome integrity. Using Xenopus egg extracts, we previously showed that replication fork collision with DPCs causes their proteolysis, followed by translesion DNA synthesis. We show here that when DPC proteolysis
Lin Deng et al.
Molecular cell, 73(5), 915-929 (2019-03-09)
DNA replication errors generate complex chromosomal rearrangements and thereby contribute to tumorigenesis and other human diseases. One mechanism that triggers these errors is mitotic entry before the completion of DNA replication. To address how mitosis might affect DNA replication, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service