Skip to Content
Merck
All Photos(1)

Key Documents

SML1259

Sigma-Aldrich

GSK2194069

≥97% (HPLC)

Synonym(s):

(S)-4-(4-(Benzofuran-5-yl)phenyl)-3-((1-(cyclopropanecarbonyl)pyrrolidin-3-yl)methyl)-1H-1,2,4-triazol-5(4H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$253.00
25 MG
$1,010.00

$253.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$253.00
25 MG
$1,010.00

About This Item

Empirical Formula (Hill Notation):
C25H24N4O3
CAS Number:
Molecular Weight:
428.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$253.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(C1CC1)N2CC[C@@H](CC(N3C4=CC=C(C5=CC=C(OC=C6)C6=C5)C=C4)=NNC3=O)C2

InChI

1S/C25H24N4O3/c30-24(18-1-2-18)28-11-9-16(15-28)13-23-26-27-25(31)29(23)21-6-3-17(4-7-21)19-5-8-22-20(14-19)10-12-32-22/h3-8,10,12,14,16,18H,1-2,9,11,13,15H2,(H,27,31)/t16-/m0/s1

InChI key

AQTPWCUIYUOEMG-INIZCTEOSA-N

Biochem/physiol Actions

GSK2194069 is a fatty acid synthase (FAS) inhibitor.
GSK2194069 is a fatty acid synthase (FAS) inhibitor. GSK2194069 is a potent and specific inhibitor of the β-ketoacyl reductase (KR) activity of hFAS with an IC50 of 7.7 ± 4.1 nM for the overall hFAS reaction. FAS is upregulated in some cancers, including prostate cancer. GSK2194069 was found to inhibit tumour growth in prostate cancer C42b cell xenografts generated in Nod-SCID-gamma mice. Cellular FAS inhibition reduced cell growth were also demonstrated in non-small-cell lung (A549) cancer cell lines with an average EC50 of 15 ± 0.5 nM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Weigt et al.
Cell chemical biology, 26(9), 1322-1331 (2019-07-08)
Human cancers require fatty acid synthase (FASN)-dependent de novo long-chain fatty acid synthesis for proliferation. FASN is therefore an attractive drug target, but fast technologies for reliable label-free cellular compound profiling are lacking. Recently, MALDI-mass spectrometry (MALDI-MS) has emerged as
Myriam Cerezo-Magaña et al.
Molecular cancer research : MCR, 19(3), 528-540 (2020-12-09)
As an adaptive response to hypoxic stress, aggressive tumors rewire their metabolic phenotype into increased malignant behavior through extracellular lipid scavenging and storage in lipid droplets (LD). However, the underlying mechanisms and potential lipid source retrieved in the hypoxic tumor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service