Skip to Content
Merck
All Photos(1)

Key Documents

C9026

Sigma-Aldrich

Caerulein

≥95% (HPLC), suitable for immunohistochemistry

Synonym(s):

Caerulein Sulfated, Cerulein, Ceruletide, [Tyr(SO3H)4]Caerulein, pGlu-Gln-Asp-Tyr(SO3H)-Thr-Gly-Trp-Met-Asp-Phe-NH2

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
$150.00
0.5 MG
$440.00
1 MG
$654.00

$150.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
0.1 MG
$150.00
0.5 MG
$440.00
1 MG
$654.00

About This Item

Empirical Formula (Hill Notation):
C58H73N13O21S2
CAS Number:
Molecular Weight:
1352.40
Beilstein:
5422487
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

$150.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Product Name

Caerulein, ≥95% (HPLC)

Quality Level

Assay

≥95% (HPLC)

form

powder

technique(s)

immunohistochemistry: suitable

color

white

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](Cc1c[nH]c2ccccc12)NC(=O)CNC(=O)C(NC(=O)[C@H](Cc3ccc(OS(O)(=O)=O)cc3)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H]4CCC(=O)N4)[C@@H](C)O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H](Cc5ccccc5)C(N)=O

InChI

1S/C58H73N13O21S2/c1-29(72)49(71-57(87)40(23-31-12-14-33(15-13-31)92-94(89,90)91)68-56(86)43(26-48(78)79)69-52(82)37(16-18-44(59)73)65-51(81)36-17-19-45(74)63-36)58(88)62-28-46(75)64-41(24-32-27-61-35-11-7-6-10-34(32)35)54(84)66-38(20-21-93-2)53(83)70-42(25-47(76)77)55(85)67-39(50(60)80)22-30-8-4-3-5-9-30/h3-15,27,29,36-43,49,61,72H,16-26,28H2,1-2H3,(H2,59,73)(H2,60,80)(H,62,88)(H,63,74)(H,64,75)(H,65,81)(H,66,84)(H,67,85)(H,68,86)(H,69,82)(H,70,83)(H,71,87)(H,76,77)(H,78,79)(H,89,90,91)/t29-,36+,37+,38+,39+,40+,41+,42+,43+,49?/m1/s1

InChI key

YRALAIOMGQZKOW-HJMGKOKZSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glp-Gln-Asp-Tyr(SO3H)-Thr-Gly-Trp-Met-Asp-Phe-NH2

Application

Caerulein has been used to induce pancreatitis in Sprague-Dawley rats, mice and other experimental animals.[1][2][3]

Biochem/physiol Actions

Caerulein is a gastro regulatory molecule with actions similar to cholecystokinin.[4] It possesses a tyrosine sulfate residue and induces smooth muscle contraction. It has gastrin-like activity.[5] Caerulein is used to study cell signaling mediated by NF-κB upregulation of proteins such as intercellular adhesion molecule-1(ICAM-1) and inflammation related molecules, such as NADPH oxidase and Janus kinase signal transducer.[6] Caerulein also regulates thermoregulation, sedation and satiety.[5]
Effects on perception of pain are conflicting. It prevents gall bladder pain, renal colic, and the pain of intermittent claudication; however, in general, it is seen as an antagonist of endogenous opioids.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acinar cell apoptosis and the origin of tubular complexes in caerulein-induced pancreatitis
Lynne E Reid
International Journal of Experimental Pathology (1999)
Shan-Ming Chen et al.
Scientific reports, 10(1), 20319-20319 (2020-11-25)
Pancreatic cancer is one of the most lethal and chemo-resistant cancers worldwide. Growing evidence supports the theory that the gut microbiota plays an essential role in modulating the host response to anti-cancer therapy. The present study aimed to explore the
Overexpression of Fas and FasL Is Associated with Infectious Complications and Severity of Experimental Severe Acute Pancreatitis by Promoting Apoptosis of Lymphocytes
Liao Pinhu
Inflammation, 37(4) (2014)
Inflammation-Induced Expression and Secretion of MicroRNA 122 Leads to Reduced Blood Levels of Kidney-Derived Erythropoietin and Anemia
Gastroenterology, 999-1010 (2016)
Enyong Dai et al.
Nature communications, 11(1), 6339-6339 (2020-12-15)
Ferroptosis is a more recently recognized form of cell death that relies on iron-mediated oxidative damage. Here, we evaluate the impact of high-iron diets or depletion of Gpx4, an antioxidant enzyme reported as an important ferroptosis suppressor, in the pancreas

Questions

1–5 of 5 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How do I store solutions of Product C9026, Caerulein?

    1 answer
    1. Solutions can be frozen at pH>7 for a least a week, and may be good for several months.

      Helpful?

  4. What is Product C9026, Caerulein, soluble in?

    1 answer
    1. It is soluble in 50 mM ammonium hydroxide at a concentration of 1 mg/mL. Note that this is information from analytical testing, and is not application based.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service