Skip to Content
Merck
All Photos(1)

Key Documents

SML1250

Sigma-Aldrich

KT203

≥98% (HPLC)

Synonym(s):

4′-[1-[[2-(Phenylmethyl)-1-piperidinyl]carbonyl]-1H-1,2,3-triazol-4-yl]-[1,1′-biphenyl]-3-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$144.00
25 MG
$451.00

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 MG
$144.00
25 MG
$451.00

About This Item

Empirical Formula (Hill Notation):
C28H26N4O3
CAS Number:
Molecular Weight:
466.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$144.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N1N=NC(C2=CC=C(C3=CC=CC(C(O)=O)=C3)C=C2)=C1)N4CCCCC4CC5=CC=CC=C5

InChI

1S/C28H26N4O3/c33-27(34)24-10-6-9-23(18-24)21-12-14-22(15-13-21)26-19-32(30-29-26)28(35)31-16-5-4-11-25(31)17-20-7-2-1-3-8-20/h1-3,6-10,12-15,18-19,25H,4-5,11,16-17H2,(H,33,34)

InChI key

SSSCOJOXPDDHOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

KT203 is a very potent, selective inhibitor of ABHD6 (IC50 = 0.82 nM). KT203 inhibits ABHD6 activity in the liver, but not the brain following ip injection in mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ku-Lung Hsu et al.
Journal of medicinal chemistry, 56(21), 8270-8279 (2013-10-25)
α/β-Hydrolase domain containing 6 (ABHD6) is a transmembrane serine hydrolase that hydrolyzes the endogenous cannabinoid 2-arachidonoylglycerol (2-AG) to regulate certain forms of cannabinoid receptor-dependent signaling in the nervous system. The full spectrum of ABHD6 metabolic activities and functions is currently

Related Content

The Cravatt group develops innovative technologies to understand enzyme roles in disease, focusing on activity-based protein profiling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service