Skip to Content
Merck
All Photos(1)

Key Documents

SML1249

Sigma-Aldrich

KT185

≥98% (HPLC)

Synonym(s):

(2-Phenylpiperidin-1-yl)(4-(3′-(piperidine-1-carbonyl)-[1,1′-biphenyl]-4-yl)-1H-1,2,3-triazol-1-yl)methanone, [4′-[1-[(2-Phenyl-1-piperidinyl)carbonyl]-1H-1,2,3-triazol-4-yl][1,1′-biphenyl]-3-yl]-1-piperidinyl-methanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$122.00
25 MG
$383.00

$122.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 MG
$122.00
25 MG
$383.00

About This Item

Empirical Formula (Hill Notation):
C32H33N5O2
CAS Number:
Molecular Weight:
519.64
UNSPSC Code:
12352200
NACRES:
NA.77

$122.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

KT185 is a very potent, selective inhibitor of ABHD6.
KT185 is a very potent, selective inhibitor of a/b-Hydrolase Domain Containing 6 (ABHD6). The compond KT185 is orally bioavailable and inhibits ABHD6 activity in the brain and liver of mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Cravatt group develops innovative technologies to understand enzyme roles in disease, focusing on activity-based protein profiling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service