Skip to Content
Merck
All Photos(1)

Key Documents

SML1180

Sigma-Aldrich

WWL229

≥98% (HPLC)

Synonym(s):

2-(3-Methoxypropyl)-1-piperidinecarboxylic acid 4-nitrophenyl ester, 4-Nitrophenyl 2-(3-methoxypropyl)piperidine-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$126.00
25 MG
$493.00

$126.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$126.00
25 MG
$493.00

About This Item

Empirical Formula (Hill Notation):
C16H22N2O5
CAS Number:
Molecular Weight:
322.36
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

$126.00


Estimated to ship onApril 11, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

oil

color

colorless to light yellow

storage temp.

2-8°C

SMILES string

O=C(OC1=CC=C([N+]([O-])=O)C=C1)N2C(CCCOC)CCCC2

InChI

1S/C16H22N2O5/c1-22-12-4-6-13-5-2-3-11-17(13)16(19)23-15-9-7-14(8-10-15)18(20)21/h7-10,13H,2-6,11-12H2,1H3

InChI key

XVBRVSAGMWRVCK-UHFFFAOYSA-N

Biochem/physiol Actions

WWL229 is a highly selective inhibitor of mouse Carboxylesterase 3 (Ces3) and human CES1 (orthologue of mCes3), serine hydrolases involved in lipolysis in addition to their activities as liver detoxification enzymes. WWL229 inhibits mCes3 with an IC50 of 10 μM, but not Ces1f, ABHD6 or other tested serine hydrolases. In a recent study, hCES1 activity was found to be increased two-fold in obese individuals and patients with type 2 diabetes compared to lean subjects, and is thought to generate surplus fatty acids that can deposit ectopically in tissues. WWL229 inhibits adipocyte basal lipolysis and promotes differentiation and lipid storage in adipocytes.
WWL229 is a highly selective inhibitor of mouse Carboxylesterase 3 (Ces3) and human CES1 (orthologue of mCes3).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Cravatt group develops innovative technologies to understand enzyme roles in disease, focusing on activity-based protein profiling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service