Skip to Content
Merck
All Photos(1)

Key Documents

SML1093

Sigma-Aldrich

Articaine hydrochloride

≥98% (HPLC)

Synonym(s):

4-Methyl-3-[[1-oxo-2-(propylamino)propyl]amino]-2-thiophenecarboxylic acid methyl ester hydrochloride, Carticaine chlorhydrate, Carticaine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$151.00

$151.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$151.00

About This Item

Empirical Formula (Hill Notation):
C13H20N2O3S · HCl
CAS Number:
Molecular Weight:
320.84
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$151.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CSC(C(OC)=O)=C1NC(C(C)NCCC)=O.Cl

InChI

1S/C13H20N2O3S.ClH/c1-5-6-14-9(3)12(16)15-10-8(2)7-19-11(10)13(17)18-4;/h7,9,14H,5-6H2,1-4H3,(H,15,16);1H

InChI key

GDWDBGSWGNEMGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Articaine hydrochloride helps to alleviate pain and blocks transmission of the pain signal.[1]
Articaine is an amide local anesthetic widely used in dentistry. Articaine acts by inhibition of nerve impulse conduction by blockade of sodium channels.
Articaine is an amide local anesthetic; sodium channel blocker.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of Lidocaine and Articaine on Neuronal Survival and Recovery.
Albalawi F, et al.
Anesthesia Progress, 65(2), 82-88 (2018)
G Batinjan et al.
International journal of oral and maxillofacial surgery, 43(12), 1503-1508 (2014-10-04)
The objective of this study was to assess the impact of low-level laser therapy on wound swelling, wound temperature changes, and oral health-related quality of life (OHRQoL) after surgical removal of impacted lower third molars. Forty patients with impacted lower

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service