Skip to Content
Merck
All Photos(1)

Key Documents

PHR1509

Supelco

Epinephrine Bitartrate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(−)-Epinephrine (+)-bitartrate salt, L-3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol D-hydrogen bitartrate salt, L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate, L-Epinephrine D-hydrogentartrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$161.00

$161.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$161.00

About This Item

Empirical Formula (Hill Notation):
C9H13NO3 · C4H6O6
CAS Number:
Molecular Weight:
333.29
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$161.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 3
traceable to Ph. Eur. A0300000
traceable to USP 1237000

API family

epinephrine

CofA

current certificate can be downloaded

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.CNC[C@H](O)c1ccc(O)c(O)c1

InChI

1S/C9H13NO3.C4H6O6/c1-10-5-9(13)6-2-3-7(11)8(12)4-6;5-1(3(7)8)2(6)4(9)10/h2-4,9-13H,5H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t9-;1-,2-/m01/s1

InChI key

YLXIPWWIOISBDD-NDAAPVSOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Epinephrine belongs to the class of drugs termed as catecholamines, which exerts its action on both alpha and beta-adrenergic receptors using a G protein-linked second messenger system.[1]

Application

Epinephrine bitartrate may be used as a pharmaceutical reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by chromatography.[2][3][4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Adrenergic receptor agonist.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0282 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of catecholamines by flow-injection analysis and high-performance liquid chromatography with chemiluminescence detection
Nalewajko E, et al.
Journal of Pharmaceutical and Biomedical Analysis, 43(5), 1673-1681 (2007)
A reversed-phase high performance liquid chromatographic method for determination of Epinephrine in pharmaceutical formulation
Mishra AK, et al
Archives of Applied Science Research, 2, 251-256 (2010)
Epinephrine
StatPearls [Internet] (2019)
Quantitative high-pressure liquid chromatographic determination of epinephrine in pharmaceutical formulations
Fu CC and Sibley MJ
Journal of Pharmaceutical Sciences, 66(3), 425-426 (1977)
George Karkoulias et al.
Cellular signalling, 66, 109446-109446 (2019-11-05)
Many aspects of neuronal development, such as neuronal survival and differentiation, are regulated by the transcription factor cAMP-response element-binding protein (CREB). We have previously reported that α2-adrenergic receptors (ARs), members of the G protein-coupled receptor (GPCR) superfamily, induce neuronal differentiation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service