Skip to Content
Merck
All Photos(1)

Key Documents

SML1069

Sigma-Aldrich

TH287 hydrochloride

≥98% (HPLC)

Synonym(s):

6-(2,3-Dichlorophenyl)-N4-methylpyrimidine-2,4-diamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$159.00

$159.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$159.00

About This Item

Empirical Formula (Hill Notation):
C11H10Cl2N4 · HCl
CAS Number:
Molecular Weight:
305.59
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$159.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

ClC1=CC=CC(C2=NC(N)=NC(NC)=C2)=C1Cl.Cl

InChI

1S/C11H10Cl2N4.ClH/c1-15-9-5-8(16-11(14)17-9)6-3-2-4-7(12)10(6)13;/h2-5H,1H3,(H3,14,15,16,17);1H

InChI key

YBLIJWDQSUDCJU-UHFFFAOYSA-N

Biochem/physiol Actions

TH287 is a potent inhibitor of human 7,8-Dihydro-8-oxoguaninetriphosphatase MTH1 (NUDT1) with an IC50 value of 0.8 nM. MTH1 hydrolyzes oxidized purine nucleoside triphosphates that might otherwise be incorporated into DNA/RNA and contribute to DNA damage. MTH1 removal of oxidized nucleotides that result from increased levels of reactive oxygen species (ROS) in fast-proliferating cancer cells helps protect cancer cells from proliferative stress and prevent cancer cell death. TH287 is considered a new target for cancer therapy. TH287 is highly selective towards MTH1, with no relevant inhibition of other members of the nudix protein family. TH287 has been shown to selectively kill a variety of cancer cell lines, but is rapidly metablized, so not as useful for in vivo studies.
TH287 is a potent inhibitor of human 7,8-Dihydro-8-oxoguaninetriphosphatase MTH1 (NUDT1).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiayu Wang et al.
Oncogene, 39(3), 603-616 (2019-09-13)
Epstein-Barr virus (EBV) immortalizes human B-lymphocytes and is implicated in the pathogenesis of lymphoid and epithelial cell malignancies. The EBV nuclear antigen (EBNA)-1 induces the accumulation of reactive oxygen species (ROS), which enables B-cell immortalization but causes oxidative DNA damage
Govindi J Samaranayake et al.
Molecular cancer therapeutics, 19(2), 432-446 (2019-11-21)
Investigations into the human 8-oxodGTPase, MutT Homolog 1 (MTH1), have risen sharply since the first-in-class MTH1 inhibitors were reported to be highly tumoricidal. However, MTH1 as a cancer therapeutic target is currently controversial because subsequently developed inhibitors did not exhibit

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service