Skip to Content
Merck
All Photos(1)

Key Documents

SML0839

Sigma-Aldrich

AGI-5198

≥98% (HPLC)

Synonym(s):

IDH-C35, N-Cycohexyl-2-(N-(3-fluorophenyl)-2-(2-methyl-1H-imidazol-1-yl)acetamido)-2-(o-tolyl)acetamide, N-[2-(Cyclohexylamino)-1-(2-methylphenyl)-2-oxoethyl]-N-(3-fluorophenyl)-2-methyl-1H-imidazole-1-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31FN4O2
CAS Number:
Molecular Weight:
462.56
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 3 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C27H31FN4O2/c1-19-9-6-7-14-24(19)26(27(34)30-22-11-4-3-5-12-22)32(23-13-8-10-21(28)17-23)25(33)18-31-16-15-29-20(31)2/h6-10,13-17,22,26H,3-5,11-12,18H2,1-2H3,(H,30,34)

InChI key

FNYGWXSATBUBER-UHFFFAOYSA-N

Application

AGI-5198 has been used as a negative control in ligand dilution series.[1]

Biochem/physiol Actions

AGI-5198 is a highly potent and selective mutant IDH1 inhibitor.
AGI-5198 is a highly potent and selective mutant isocitrate dehydrogenase 1 (IDH1) inhibitor.
In human chondrosarcoma cell line JJ012, AGI-5198 promotes apoptosis and G2/M cell cycle arrest.[2] It also prevents the generation of colony in chondrosarcoma cell lines. AGI-5198 hinders the multiplication of cell and induces demethylation of H3K9me3 and H3K27me3.[3]

Other Notes

AGI-5198 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the AGI-5198 probe summary on the Chemical Probes Portal website.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luyuan Li et al.
PloS one, 10(9), e0133813-e0133813 (2015-09-15)
Chondrosarcomas are malignant bone tumors that produce cartilaginous matrix. Mutations in isocitrate dehydrogenase enzymes (IDH1/2) were recently described in several cancers including chondrosarcomas. The IDH1 inhibitor AGI-5198 abrogates the ability of mutant IDH1 to produce the oncometabolite D-2 hydroxyglutarate (D-2HG)
Near-native, site-specific and purification-free protein labeling for quantitative protein interaction analysis by MicroScale Thermophoresis.
Bartoschik T, et al.
Scientific Reports, 8(1), 4977-4977 (2018)
Frontiers in Clinical Drug Research, 495-495 (2016)
Geon-Hee Kim et al.
International journal of molecular sciences, 20(11) (2019-06-04)
The R132H mutation in isocitrate dehydrogenase 1 (IDH1R132H) is commonly observed and associated with better survival in glioblastoma multiforme (GBM), a malignant brain tumor. However, the functional role of IDH1R132H as a molecular target for GBM treatment is not completely
Kancharana Bala Bhaskara Rao et al.
FEBS letters, 593(16), 2177-2193 (2019-06-19)
Isocitrate dehydrogenases (IDHs) are metabolic enzymes that are mutated in several cancers, resulting in overproduction of d-2-hydroxyglutarate (D-2HG). However, the signalling pathways and factors that regulate mutant IDHs or their metabolites remain elusive. Here, we report that in synchronized cells

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service