Skip to Content
Merck

Skip To

SML0809

LE 135

≥98% (HPLC)

Synonym(s):

4-(7,8,9,10-Tetrahydro-5,7,7,10,10-pentamethyl-5H-benzo[e]naphtho[2,3-b][1,4]diazepin-13-yl)-benzoic acid, LE-135

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
$131.00
$111.35

About This Item

Empirical Formula (Hill Notation):
C29H30N2O2
CAS Number:
Molecular Weight:
438.56
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

$111.35

List Price$131.00Save 15%
PRICE WITHOUT NATIONAL TAXES
Web-Only Promotion

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

N1(c2c(cc5c(c2)C(CCC5(C)C)(C)C)N=C(c4c1cccc4)c3ccc(cc3)C(=O)O)C

InChI

1S/C29H30N2O2/c1-28(2)14-15-29(3,4)22-17-25-23(16-21(22)28)30-26(18-10-12-19(13-11-18)27(32)33)20-8-6-7-9-24(20)31(25)5/h6-13,16-17H,14-15H2,1-5H3,(H,32,33)

InChI key

YZZAIQOVMHVWBS-UHFFFAOYSA-N

Application

LE 135 has been used for in vitro islet treatment.

Biochem/physiol Actions

LE135 is a retinoic acid receptor (RAR) antagonist with selectivity for RARβ (Ki = 220 nM) over RARα (Ki = 1.4 μM).
LE135 is a retinoic acid receptor (RAR) antagonist with selectivity for RARβ (Ki = 220 nM) over RARα (Ki = 1.4 μM). LE135 inhibits retinoic acid-induced transcriptional activation of RARβ (>70% inhibition at 10 μM), but not RARα, RARγ or retinoid X receptor α (RXRα). There is high interest in retinoic acid receptors for cancer and for differentiation studies. LE135 has been shown to inhibit retinoid Am80-induced differentiation of human promyelocytic leukemia HL-60 cells with an IC50 value of 0.2 μM. LE135 has been used to study the role of a retinoic acid receptor-β (RARβ) on the differentiation of mesenchymal stem cells, and was found to inhibit the neuronal differentiation promoting effects of all-trans retinoic acid (ATRA) on mesenchymal stem cells.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2170SML1753SML2233
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

solubility

DMSO: 15 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear (warmed)

color

yellow to orange

color

yellow to orange

color

white to beige

color

white to beige


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters