Skip to Content
Merck
All Photos(1)

Key Documents

SML0708

Sigma-Aldrich

BMS-182874 hydrochloride

≥98% (HPLC)

Synonym(s):

5-(Dimethylamino)-N-(3,4-dimethyl-5-isoxazolyl)-1-naphthalenesulfonamide hydrochloride, BMS182874, N-(3,4-Dimethyl-5-isoxazolyl)-5-(dimethylamino)-1-naphthalenesulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3O3S · HCl
CAS Number:
Molecular Weight:
381.88
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

InChI

1S/C17H19N3O3S.ClH/c1-11-12(2)18-23-17(11)19-24(21,22)16-10-6-7-13-14(16)8-5-9-15(13)20(3)4;/h5-10,19H,1-4H3;1H

InChI key

UZZFRNZGYDOBAO-UHFFFAOYSA-N

Biochem/physiol Actions

BMS-182874 is a potent selective nonpeptide endothelin ETA receptor antagonist. BMS-182874 is 1000-fold selective for ETA over ETA B receptors with a Ki of 48 nM for ETA receptors vs >50 μM for ETB.

Features and Benefits

This compound is featured on the Endothelin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service