Skip to Content
Merck
All Photos(1)

Key Documents

SML0424

Sigma-Aldrich

ST247

≥98% (HPLC)

Synonym(s):

3-[[[4-(Hexylamino)-2-methoxyphenyl]amino]sulfonyl]-2-thiophenecarboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$290.00

$290.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$290.00

About This Item

Empirical Formula (Hill Notation):
C19H26N2O5S2
CAS Number:
Molecular Weight:
426.55
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$290.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL (clear solution)

storage temp.

2-8°C

SMILES string

CCCCCCNc1ccc(NS(=O)(=O)c2ccsc2C(=O)OC)c(OC)c1

InChI

1S/C19H26N2O5S2/c1-4-5-6-7-11-20-14-8-9-15(16(13-14)25-2)21-28(23,24)17-10-12-27-18(17)19(22)26-3/h8-10,12-13,20-21H,4-7,11H2,1-3H3

InChI key

MRWCGEOYGFZZCJ-UHFFFAOYSA-N

Biochem/physiol Actions

ST247 is a PPAR β/δ inverse agonist.
ST247 is a PPAR β/δ selective inverse agonist. The compound is based on the structure of GSK0660, and is 10-fold more potent in blocking the transcription of endogenous target genes: ST247 IC50 = 19 nm vs. GSK0660 IC50 = 210 nM.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Yu et al.
The Journal of clinical endocrinology and metabolism, 105(9) (2020-07-01)
Implantation is a reproductive bottleneck in women, regulated by fluctuations in ovarian steroid hormone concentrations. However, other nuclear receptor ligands are modifiers of endometrial differentiation leading to successful pregnancy. In the present study we analyzed the effects of peroxisome-proliferator-activated receptor
Christina Blücher et al.
Molecular cancer research : MCR, 18(12), 1849-1862 (2020-08-30)
Recent studies indicate that adipose tissue in obesity promotes breast cancer progression by secreting protumorigenic chemokines, growth factors, and fatty acids. However, the detailed mechanisms by which hypertrophic adipose tissue influences breast cancer cells are still not well understood. Here

Articles

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

We offer many products related to non-steroid nuclear receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service