Skip to Content
Merck
All Photos(1)

Key Documents

SML0290

Sigma-Aldrich

MLS000389544

≥98% (HPLC)

Synonym(s):

4-(Methylsulfonyl)-3-nitro-benzoic acid 2-oxo-2-(tricyclo[3.3.1.13,7]dec-1-ylamino)ethyl ester, MLS-389544, [2-(1-Adamantylamino)-2-oxoethyl] 4-methylsulfonyl-3-nitrobenzoate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$168.00
25 MG
$673.00

$168.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$168.00
25 MG
$673.00

About This Item

Empirical Formula (Hill Notation):
C20H24N2O7S
CAS Number:
Molecular Weight:
436.48
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$168.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: ≥5 mg/mL (warmed)

storage temp.

−20°C

SMILES string

CS(=O)(=O)c1ccc(cc1[N+]([O-])=O)C(=O)OCC(=O)NC23CC4CC(CC(C4)C2)C3

InChI

1S/C20H24N2O7S/c1-30(27,28)17-3-2-15(7-16(17)22(25)26)19(24)29-11-18(23)21-20-8-12-4-13(9-20)6-14(5-12)10-20/h2-3,7,12-14H,4-6,8-11H2,1H3,(H,21,23)

InChI key

BTXTVUJWNPMQBT-UHFFFAOYSA-N

Biochem/physiol Actions

MLS000389544 is a selective and potent methylsulfonylnitrobenzoate thyroid hormone receptor β (TRβ) antagonist that blocks the association of TRβ with steroid receptor coactivator 2 (SRC2).
MLS000389544 is a selective and potent methylsulfonylnitrobenzoate thyroid hormone receptor β (TRβ) antagonist that blocks the association of TRβ with steroid receptor coactivator 2 (SRC2). It effectively block TR-mediated gene transcription. MLS000389544 is selective for TR over other NR family members. It appears to act by irreversible mechanisms and target cysteine residues in the coactivator binding pocket.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to non-steroid nuclear receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service