Skip to Content
Merck
All Photos(1)

Key Documents

SML0287

Sigma-Aldrich

680C91

≥98% (HPLC)

Synonym(s):

6-Fluoro-3-[(1E)-2-(3-pyridinyl)ethenyl)-1H-indole

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$138.00
25 MG
$554.00

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$138.00
25 MG
$554.00

About This Item

Empirical Formula (Hill Notation):
C15H11FN2
CAS Number:
Molecular Weight:
238.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$138.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

Fc1ccc2c(\C=C\c3cccnc3)c[nH]c2c1

InChI

1S/C15H11FN2/c16-13-5-6-14-12(10-18-15(14)8-13)4-3-11-2-1-7-17-9-11/h1-10,18H/b4-3+

InChI key

YBSDQTBCNYWBMX-ONEGZZNKSA-N

Application

680C91 has been used:
  • as a tryptophan 2,3 dioxygenase (TDO) inhibitor to study its effects on the pigmentation in Doryteuthis pealeii embryos[1]
  • as a TDO inhibitor to study its effects on esophageal squamous cell carcinoma in xenograft tumor assay[2]
  • as a tryptophan 2,3 dioxygenase 2 (TDO2) inhibitor to study its effects on toxic fragment formation in human embryonic kidney cells[3]

Biochem/physiol Actions

680C91 is a potent inhibitor of the enzyme tryptophan 2,3-dioxygenase (TDO), which directs the conversion of trypophan to kynurenin. Kynurenin has recently been identified as an endogenous lignd of the arylhydrocarbon receptor (AHR). TDO is highly expressed in glioma cells, and contributes to AHR-mediated glioma cell survival and suppression of anti-tumor immune responses.
680C91 is a potent inhibitor of the enzyme tryptophan 2,3-dioxygenase (TDO).
680C91 shows no inhibitory effects against monoamine oxidase A and B, indoleamine 2,3-dioxygenase and serotonin (5-HT) uptake, and 5-HT1A,1D,2A, and 2C receptors.[4]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Paccosi et al.
Journal of cancer research and clinical oncology, 146(12), 3155-3163 (2020-08-11)
Indoleamine 2,3-dioxygenase-1 (IDO1) and more recently, tryptophan 2,3-dioxygenase (TDO), are tryptophan-catabolizing enzymes with immunoregulatory properties in cancer. IDO1 is more expressed than TDO in many tumours including melanomas; however, IDO inhibitors did not give expected results in clinical trials, highlighting
Karthyayani Rajamani et al.
Neuropharmacology, 117, 434-446 (2017-02-23)
Spinocerebellar ataxia type 3 or Machado-Joseph disease (SCA3/MJD) is characterized by the repetition of a CAG codon in the ataxin-3 gene (ATXN3), which leads to the formation of an elongated mutant ATXN3 protein that can neither be denatured nor undergo
Carlo Breda et al.
Proceedings of the National Academy of Sciences of the United States of America, 113(19), 5435-5440 (2016-04-27)
Metabolites of the kynurenine pathway (KP) of tryptophan (TRP) degradation have been closely linked to the pathogenesis of several neurodegenerative disorders. Recent work has highlighted the therapeutic potential of inhibiting two critical regulatory enzymes in this pathway-kynurenine-3-monooxygenase (KMO) and tryptophan-2,3-dioxygenase
Tryptophan 2, 3-dioxygenase 2 controls M2 macrophages polarization to promote esophageal squamous cell carcinoma progression via AKT/GSK3 beta IL-8 signaling pathway
Zhao Y, et al.
Acta pharmaceutica Sinica. B (2021)
M Salter et al.
Biochemical pharmacology, 49(10), 1435-1442 (1995-05-17)
The effects of a novel inhibitor 680C91 ((E)-6-fluoro-3-[2-(3- pyridyl)vinyl]-1H-indole) of the key enzyme of tryptophan catabolism tryptophan 2,3-dioxygenase (TDO) (EC 1.13.11.11), were examined on tryptophan catabolism in vitro and in vivo and on brain levels of tryptophan, serotonin (5-HT) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service