Skip to Content
Merck
All Photos(1)

Key Documents

SML0225

Sigma-Aldrich

LY255582

≥98% (HPLC)

Synonym(s):

(3R,4R)-3,4-Dimethyl-1-[(3S)-3-hydroxy-3-cyclohexyl-propyl]-4-(3-hydroxyphenyl)piperidine, (aS,3R,4R)-a-Cyclohexyl-4-(3-hydroxyphenyl)-3,4-dimethyl-1-piperidinepropanol, LY-255582

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$153.00
25 MG
$606.00

$153.00


Estimated to ship onApril 25, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$153.00
25 MG
$606.00

About This Item

Empirical Formula (Hill Notation):
C22H35NO2
CAS Number:
Molecular Weight:
345.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$153.00


Estimated to ship onApril 25, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: ≥15 mg/mL

originator

Eli Lilly

storage temp.

2-8°C

SMILES string

C[C@H]1CN(CC[C@H](O)C2CCCCC2)CC[C@@]1(C)c3cccc(O)c3

InChI

1S/C22H35NO2/c1-17-16-23(13-11-21(25)18-7-4-3-5-8-18)14-12-22(17,2)19-9-6-10-20(24)15-19/h6,9-10,15,17-18,21,24-25H,3-5,7-8,11-14,16H2,1-2H3/t17-,21-,22+/m0/s1

InChI key

LVVHEFJXPXAUDD-BULFRSBZSA-N

Application

LY255582 may be used in opioid receptor-mediated cell signaling studies.

Biochem/physiol Actions

LY255582 inhibits the diet-associated increases in mesolimbic dopamine levels and reduces the consumption of highly-palatable food intake.[1]
LY255582 is a centrally active opioid receptor antagonist
LY255582 is a centrally active opioid receptor antagonist (defined as an inverse agonist in 2011 JPET paper) that inhibited weight gain in obese Zucker rats over 30 days. It is more that 5-fold selective for mu opioid receptor compared to kappa opioid receptors and 13-fold selective over delta opioid receptors.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allison E Sahr et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 295(2), R463-R471 (2008-06-06)
An analog of the trans-3,4-dimethyl-4-(3-hydroxyphenyl)piperidine series (LY255582) exhibits high in vitro binding affinity and antagonist potency for the mu-, delta-, and kappa-opioid receptors. In vivo, LY255582 exhibits potent effects in reducing food intake and body weight in several rodent models

Articles

We offers many products related to opioid receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service