Skip to Content
Merck
All Photos(2)

Key Documents

SMB00915

Sigma-Aldrich

BAA473 (11-oxo-12S-hydroxy lithocholic acid methyl ester)

≥95% (HPLC)

Synonym(s):

BAA473 (11-oxo-12S-hydroxy lithocholic acid methyl ester), (3α,5β,12β)-3,12-dihydroxy-11-oxo-cholan-24-oic-acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$447.00

$447.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$447.00

About This Item

Empirical Formula (Hill Notation):
C25H40O5
CAS Number:
Molecular Weight:
420.58
UNSPSC Code:
41141802
NACRES:
NA.25

$447.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

mp

153.5—154.5 °C

solubility

chloroform: 20 mg/mL, colorless to light yellow

storage temp.

2-8°C

SMILES string

O=C(CCC(C1CCC2C3CCC4CC(CCC4(C3C(C(C12C)O)=O)C)O)C)OC

InChI

1S/C25H40O5/c1-14(5-10-20(27)30-4)18-8-9-19-17-7-6-15-13-16(26)11-12-24(15,2)21(17)22(28)23(29)25(18,19)3/h14-19,21,23,26,29H,5-13H2,1-4H3

InChI key

RYGJYUKSMUSEMY-UHFFFAOYSA-N

General description

BAA473 (11-oxo-12S-hydroxy lithocholic acid methyl ester) is a more potent analog of microbiome metabolite BAA485 (12-oxolithocholic acid) that activates the Pyrin inflammasome in myeloid and intestinal epithelial cells. BAA473 potently induces secretion of IL1β and IL18 in LPS-primed PBMCs and THP-1 cells.
Bile acids, natural steroid amphipathic compounds formed through cholesterol oxidation in the liver, play a crucial role in regulating lipid metabolism. They facilitate the digestion, absorption, and transport of lipids, lipid-soluble vitamins, and related molecules in the gastrointestinal tract. These bile acids also impact various aspects of cholesterol, glucose, and energy balance.

Application

BAA473 (11-oxo-12S-hydroxy lithocholic acid methyl ester) finds application in metabolomics, biochemical and microbiome research.

Features and Benefits

  • Can be used in Metabolomics and Biochemical research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irina Alimov et al.
The Journal of biological chemistry, 294(10), 3359-3366 (2019-01-17)
Bile acids are critical metabolites in the gastrointestinal tract and contribute to maintaining intestinal immune homeostasis through cross-talk with the gut microbiota. The conversion of bile acids by the gut microbiome is now recognized as a factor affecting both host

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service