Skip to Content
Merck
All Photos(1)

Key Documents

SML0056

Sigma-Aldrich

Cefotiam dihydrochloride hydrate

≥98% (HPLC)

Synonym(s):

(6R,7R)-7-[[2-(2-Amino-4-thiazolyl)acetyl]amino]-3-[[[1-[2-(dimethylamino)ethyl]-1H-tetrazol-5-yl]-thio]methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid dihydrochloride hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$108.00
50 MG
$336.00

$108.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$108.00
50 MG
$336.00

About This Item

Empirical Formula (Hill Notation):
C18H23N9O4S3 · 2HCl · xH2O
CAS Number:
Molecular Weight:
598.55 (anhydrous basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$108.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

H2O: ≥22 M

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1)CC(=O)N[C@H]2[C@H]3SCC(=C(N3C2=O)C(=O)O)CSc4[n](nnn4)CCN(C)C)N.[Cl-].[Cl-].[H+].[H+]

InChI

1S/C18H23N9O4S3.2ClH/c1-25(2)3-4-26-18(22-23-24-26)34-7-9-6-32-15-12(14(29)27(15)13(9)16(30)31)21-11(28)5-10-8-33-17(19)20-10;;/h8,12,15H,3-7H2,1-2H3,(H2,19,20)(H,21,28)(H,30,31);2*1H/t12-,15-;;/m1../s1

InChI key

BWRRTAXZCKVRON-DGPOFWGLSA-N

General description

Chemical structure: ß-lactam

Application

Cefotiam dihydrochloride hydrate is used as broad-spectrum antibiotic.

Biochem/physiol Actions

Cefotiam is a second generation cephalosporin antibiotic which has been used as a prophylatic antibiotic for MRSA.
Cefotiam, a derivative of cephalosporin, exhibits broad-spectrum anti-bacterial activity. It inhibits the cross-linking stage of peptidoglycan and blocks the synthesis of bacterial cell wall. It is resistant to β-lactamases.[1][2]
Second generation cephalosporin, prophylatic antibiotic for MRSA

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rainer Müller et al.
Arzneimittel-Forschung, 53(2), 126-132 (2003-03-20)
The efficacy of the perioperative short-term prophylaxis with cefotiam (CAS 66309-69-1) and cefuroxime axetil (CAS 64544-07-6) was analysed by the assessment of the pharmacological kinetics in the serum and the tonsil tissue in 50 patients with recurrent tonsillitis. Twenty-four patients
J R Lentino et al.
Antimicrobial agents and chemotherapy, 25(6), 778-780 (1984-06-01)
Cefotiam was evaluated by a comparative open-label randomized trial with cephalothin in the therapy of skin and soft tissue infections in 39 patients. The most common organism isolated was Staphylococcus aureus (78%). We established evidence of primary infection with gram-negative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service