Skip to Content
Merck
All Photos(1)

Key Documents

SML0025

Sigma-Aldrich

Safinamide mesylate salt

≥98% (HPLC)

Synonym(s):

(S)-(+)-2-[[4-(3-Fluorobenzoxy)benzyl]amino]propanamide; (S)-2-[[4-[(3-Fluorobenzyl)oxy]benzyl]amino]propanamide; FCE-26743; PNU-151774E, NW-1015;

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$166.00
25 MG
$648.00

$166.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$166.00
25 MG
$648.00

About This Item

Empirical Formula (Hill Notation):
C17H19FN2O2 · xCH4O3S
CAS Number:
Molecular Weight:
302.34 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$166.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +9.5 to +14°, c = 1 (95% acetic acid)

color

white to tan

solubility

H2O: ≥15 mg/mL

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.C[C@H](NCc1ccc(OCc2cccc(F)c2)cc1)C(N)=O

InChI

1S/C17H19FN2O2.CH4O3S/c1-12(17(19)21)20-10-13-5-7-16(8-6-13)22-11-14-3-2-4-15(18)9-14;1-5(2,3)4/h2-9,12,20H,10-11H2,1H3,(H2,19,21);1H3,(H,2,3,4)/t12-;/m0./s1

InChI key

YKOCHIUQOBQIAC-YDALLXLXSA-N

Gene Information

human ... MAOB(4129)

Looking for similar products? Visit Product Comparison Guide

Application

Safinamide mesylate salt has been used as a reference drug to study its inhibitory effect on human monoamine oxidases (hMAO-A and hMAO-B).[1]
Safinamide mesylate salt may be used in cell signaling studies.

Biochem/physiol Actions

Safinamide is a highly selective and reversible monoamine oxidase type B (MAO-B) inhibitor that increases neostriatal dopamine concentration. In addition, safinamide is voltage-dependent sodium and calcium channel blocker. It appears to bind to the batrachotoxin-sensitive site 2 of the voltage-sensitive sodium channels. Safinamide blocks N and L-type calcium channels and inhibits glutamate and aspartate release from synaptic terminals.
Safinamide is a sodium and calcium channel blocker that is also a selective and reversible inhibitor of monoamine oxidase type B (MAO-B); anticonvulsant
Safinamide mesylate prevents dopamine reuptake and is effective in the treatment of epilepsy and Parkinson′s disease.

Features and Benefits

This compound is featured on the Calcium Channels and Dopamine and Norepinephrine Metabolism pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of human monoamine oxidase: biological and molecular modeling studies on selected natural flavonoids
Carradori S, et al.
Journal of Agricultural and Food Chemistry, 64(47), 9004-9011 (2016)
Se Eun Park et al.
Journal of agricultural and food chemistry, 68(39), 10719-10729 (2020-09-02)
Luteolin, a flavonoid widely distributed in the plant kingdom, contains two benzene rings and hydroxyl groups, and this structural specificity contributes to its diverse biological activities. However, no previous studies have simultaneously investigated the therapeutic potency of luteolin isolated from
Diana Duarte et al.
International journal of molecular sciences, 22(14) (2021-07-25)
Several central nervous system (CNS) drugs exhibit potent anti-cancer activities. This study aimed to design a novel model of combination that combines different CNS agents and antineoplastic drugs (5-fluorouracil (5-FU) and paclitaxel (PTX)) for colorectal and breast cancer therapy, respectively.

Articles

Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials.

We offers many products related to calcium channels for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service