Skip to Content
Merck
All Photos(1)

Key Documents

SMB01078

Sigma-Aldrich

Inulicin

≥90% (LC/MS-ELSD)

Synonym(s):

1-O-Acetylbritannilactone, Britannilactone 1-O-acetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$459.00

$459.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$459.00

About This Item

Empirical Formula (Hill Notation):
C17H24O5
CAS Number:
Molecular Weight:
308.37
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

$459.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

308.37

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@H]2[C@H]([C@@H](C(=C(C2)C)[C@H](CCCOC(=O)C)C)O)C(=C)C1=O

InChI

1S/C17H24O5/c1-9(6-5-7-21-12(4)18)14-10(2)8-13-15(16(14)19)11(3)17(20)22-13/h9,13,15-16,19H,3,5-8H2,1-2,4H3/t9-,13+,15+,16+/m0/s1

InChI key

QKUFZFLZBUSEHN-CZLJMHDISA-N

General description

Inulicin, a sesquiterpene lactone, is a natural bioactive compound commonly sourced from plants like Pentanema britannicum and Inula japonica. Current research indicates that this plant-derived metabolite may possess inhibitory properties, showcasing a range of biological activities, including anti-inflammatory, cardioprotective, antiangiogenic, and anticancer activities.

Application

Inulicin is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

Inulicin exhibits anticancer properties by inhibiting angiogenesis and lung cancer cell proliferation, potentially through the regulation of VEGFR-Src-FAK signaling. When combined with gemcitabine, it promotes strong apoptosis in lung cancer cells. Additionally, Inulicin shows promise in the treatment of various cardiovascular diseases, including chronic ischemia, by modulating VEGF signaling and angiogenesis. It also serves as a potent inhibitor of LPS-induced inflammatory responses in vascular smooth muscle cells, acting through the blockade of NF-kappaB activity and the suppression of the inflammatory gene COX-2. Furthermore, Inulicin displays potential as a natural skin-lightening agent, reducing melanogenesis by suppressing tyrosinase expression via ERK and Akt signaling pathways.

Features and Benefits

  • Suitable for Biochemical and Biomedical research
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L F Belova et al.
Farmakologiia i toksikologiia, 44(4), 463-467 (1981-07-01)
Inulicin is a sesquiterpene lactone obtained from Inula Japonica L. (Compositae) possesses a fairly wide spectrum of pharmacological action. In doses from 5 to 60 mg/kg it exerts a certain stimulant action on the central nervous system and smooth muscles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service