Skip to Content
Merck
All Photos(2)

Key Documents

SMB00472

Sigma-Aldrich

Palmatine chloride

≥98% (HPLC)

Synonym(s):

Berbinium, Dibenzo[a,g]quinolizinium,5,6-dihydro-2,3,9,10-tetramethoxychloride, Fibrauretin chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$360.00

$360.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$360.00

About This Item

Empirical Formula (Hill Notation):
C21H22ClNO4
CAS Number:
Molecular Weight:
387.86
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$360.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

shipped in

wet ice

storage temp.

−20°C

SMILES string

COC1=CC2=C(C=C1OC)CC[N+]3=CC4=C(OC)C(OC)=CC=C4C=C32.[Cl-]

InChI

1S/C21H22NO4.ClH/c1-23-18-6-5-13-9-17-15-11-20(25-3)19(24-2)10-14(15)7-8-22(17)12-16(13)21(18)26-4;/h5-6,9-12H,7-8H2,1-4H3;1H/q+1;/p-1

InChI key

RLQYRXCUPVKSAW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Palmatineis an isoquinoline alkaloid. Palmatine chloride (PMTCl) is a protoberberine isoquinoline alkaloid with no ketone group and a soluble drug form of palmatine. It has low hygroscopic stability because of chloride ions present in its structure. PMTCl is a natural compound found in the rhizomes of Coptischinensis.

Application

Palmatine chloride has been used to test its effects as an inhibitor of soluble epoxide hydrolase (sEH) in vitro.

Biochem/physiol Actions

Palmatine chloride acts as a microbial collagenase inhibitor and a potential anti-infective agent. It possesses many other pharmacological properties such as antioxidant, anti-inflammatory, anticancer, anti-viral, and neuroprotective.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Georgiana Nitulescu et al.
Life (Basel, Switzerland), 12(12) (2022-12-24)
Bacterial virulence factors are mediating bacterial pathogenesis and infectivity. Collagenases are virulence factors secreted by several bacterial stains, such as Clostridium, Bacillus, Vibrio and Pseudomonas. These enzymes are among the most efficient degraders of collagen, playing a crucial role in
Improving hygroscopic stability of palmatine chloride by forming a pharmaceutical salt cocrystal of palmatine chloride-gallic acid with neutral molecule
Zhang YN, et al.,
Journal of drug delivery science and technology, 66, 102839-102839 (2021)
Jang Hoon Kim et al.
Current issues in molecular biology, 44(9), 4282-4289 (2022-09-23)
The quaternary isoquinoline alkaloids of palmatine (1), berberine (2), and jatrorrhizine (3) were evaluated in terms of their ability to inhibit soluble epoxide hydrolase (sEH). They had similar inhibitory activities, with IC50 values of 29.6 ± 0.5, 33.4 ± 0.8

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service