Skip to Content
Merck
All Photos(1)

Key Documents

SMB01059

Sigma-Aldrich

Coatline B

≥90% (LC/MS-ELSD)

Synonym(s):

(2R)-3-(3,4-Dihydroxyphenyl)-1-(3-β-D-glucopyranosyl-2,4-dihydroxyphenyl)-2-hydroxy-1-propanone (ACI)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$466.00

$466.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$466.00

About This Item

Empirical Formula (Hill Notation):
C21H24O11
CAS Number:
Molecular Weight:
452.41
UNSPSC Code:
12352205
NACRES:
NA.25

$466.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

452.41

solubility

water: slightly soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1[C@@H]([C@H]([C@@H]([C@H]([C@@H]1c2c(c(ccc2O)C(=O)[C@H](O)Cc3cc(c(cc3)O)O)O)O)O)O)CO

InChI key

LNVWGAALAQOWHF-IUOSPEFCSA-N

General description

Coatline B, a dihydroxychalcone, is a natural product commonly found in plants such as Eysenhardtia polystachya and Cassia nomame. It serves as a secondary metabolite of these plants, boasting antidiabetic, antibacterial, and antioxidant properties. With unique characteristics like photoluminescence, Coatline B holds significance in various research areas, including biomarker and biosensor development, compound screening libraries, metabolite profiling, and phytochemical and biochemical research.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service