Skip to Content
Merck
All Photos(1)

Key Documents

SMB01075

Sigma-Aldrich

Myrtucommulone B

≥90% (LC/MS-ELSD)

Synonym(s):

6,8-dihydroxy-2,2,4,4-tetramethyl-5-(2-methylpropanoyl)-9-propan-2-yl-9H-xanthene-1,3-dione

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$517.00

$517.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$517.00

About This Item

Empirical Formula (Hill Notation):
C24H30O6
CAS Number:
Molecular Weight:
414.49
UNSPSC Code:
12352205
NACRES:
NA.25

$517.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

414.49

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1c2c(c(cc(c2C(=O)C(C)C)O)O)C(C3=C1C(C(=O)C(C3=O)(C)C)(C)C)C(C)C

InChI

1S/C24H30O6/c1-10(2)14-15-12(25)9-13(26)16(18(27)11(3)4)19(15)30-21-17(14)20(28)23(5,6)22(29)24(21,7)8/h9-11,14,25-26H,1-8H3

InChI key

AYAUOXWJIWVPSO-UHFFFAOYSA-N

General description

Myrtucommulone B, a polyprenylated acylphloroglucinol, is a naturally occurring substance typically extracted from Myrtus communis (Myrtaceae). Previous studies have indicated that this natural compound derived from plants possesses a range of biological activities, including antibacterial, anti-inflammatory, and antioxidant properties

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Biochem/physiol Actions

According to the existing research, Myrtucommulone B exhibited strong inhibition of sEH (soluble epoxide hydrolase) activity, suggesting its potential role in regulating lipid metabolism and inflammation[1]. Further, it also showed significant inhibition of α-glucosidase activity suggesting its potential as antidiabetic and antibacterial agents[2].

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New ?-Glucosidase Inhibitors and Antibacterial Compounds fromMyrtus communis L.
Shaheen F, et al.
European Journal of Organic Chemistry, 2006, 2371-2377 (2006)
Pham Ngoc Khanh et al.
Fitoterapia, 109, 39-44 (2015-11-10)
Phytochemical analysis of the leaves and stems of Callistemon citrinus (Curtis) Skeels led to the isolation of two new alkylphloroglucinols, gallomyrtucommulone E and F (1 and 2), along with four other known alkylphloroglucinol derivatives, gallomyrtucommulone A (3), endoperoxide G3 (4)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service