Skip to Content
Merck
All Photos(1)

Key Documents

SMB01015

Sigma-Aldrich

Paederosidic acid g-lactone

≥90% (LC/MS-ELSD)

Synonym(s):

1H-2,6-Dioxacyclopent[cd]indene, carbonothioic acid deriv. (ZCI), Paederoside, Pederoside

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$466.00

$466.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$466.00

About This Item

Empirical Formula (Hill Notation):
C18H22O11S
CAS Number:
Molecular Weight:
446.43
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

$466.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

446.43

solubility

water: soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

S(C)C(=O)OCC1=C[C@@H]2OC(=O)C3=CO[C@H]([C@H]1[C@@H]32)O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)CO

InChI

1S/C18H22O11S/c1-30-18(24)26-4-6-2-8-11-7(15(23)27-8)5-25-16(10(6)11)29-17-14(22)13(21)12(20)9(3-19)28-17/h2,5,8-14,16-17,19-22H,3-4H2,1H3/t8-,9+,10+,11-,12+,13-,14+,16-,17-/m0/s1

InChI key

OJISWUQNQQWEND-FCVLBCLDSA-N

General description

Paederosidic acid g-lactone, also known as Paederoside, is a monoterpene iridoid glycoside that is commonly found in the Paederia Sp., Plocama calabrica, Pseudomegoura nipponica, and Asperula erythrosa. This plant metabolite exhibits various biological activities, anti-inflammatory, antioxidant, antimicrobial, anticancer, and neuroprotective activities.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service