Skip to Content
Merck
All Photos(1)

Documents

SMB00386

Sigma-Aldrich

Tea Tree Oil

Synonym(s):

Tea tree oil, Melaleuca alternifolia oil, Melasol, Melaleuca alternifolia

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211

biological source

Melaleuca alternifolia

form

liquid

refractive index

n20/D 1.478 (lit.)

bp

165 °C (lit.)

density

0.898 g/mL at 25 °C (lit.)

lipid type

oils

Looking for similar products? Visit Product Comparison Guide

General description

Tea Tree Oil is an oil found in the leaves of Melaleuca alternifolia. This essential oil is composed of terpene hydrocarbon, mainly monoterpenes and sesquiterpenes. Tea tree oil is volatile and slightly soluble in water.

Application

Tea Tree Oil has been used to induce stress response in Saccharomyces cerevisiae.

Biochem/physiol Actions

Tea Tree Oil possesses antimicrobial, anti scabies, and antifungal properties. It is commonly used in cosmeceuticals. Tea tree oil is also used to promote wound healing and to decrease periodic breast pain. Higher dose of tea tree oil is considered toxic and might result in skin allergy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.2 °F

Flash Point(C)

64 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tryptophan recovers sensitivity to cell membrane stress in Saccharomyces cerevisiae
Ikui A, et al.
bioRxiv, 44(5), 344366-344366 (2018)
David Larson et al.
Dermatitis : contact, atopic, occupational, drug, 23(1), 48-49 (2012-06-02)
Tea tree oil is an increasingly popular ingredient in a variety of household and cosmetic products, including shampoos, massage oils, skin and nail creams, and laundry detergents. Known for its potential antiseptic properties, it has been shown to be active
K A Hammer et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 44(5), 616-625 (2005-10-26)
The essential oil of Melaleuca alternifolia, also known as tea tree or melaleuca oil, is widely available and has been investigated as an alternative antimicrobial, anti-inflammatory and anti-cancer agent. While these properties are increasingly well characterised, relatively limited data are
Lea Schroeder et al.
PloS one, 14(3), e0199484-e0199484 (2019-03-12)
Sodium dodecyl sulfate is a detergent that disrupts cell membranes, activates cell wall integrity signaling and restricts cell growth in Saccharomyces cerevisiae. However, the underlying mechanism of how sodium dodecyl sulfate inhibits cell growth is not fully understood. Previously, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service