Skip to Content
Merck
All Photos(1)

Key Documents

24245

Sigma-Aldrich

Oil of turpentine

purified

Synonym(s):

Fir oil, Turpentine oil, Oil of turpentine, Pine oil, Turpentine

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$217.00
2.5 L
$228.00

$217.00


Estimated to ship onApril 02, 2025


Request a Bulk Order

Select a Size

Change View
1 L
$217.00
2.5 L
$228.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22
bp:
153-175 °C (lit.)
vapor pressure:
4 mmHg ( −6.7 °C)

$217.00


Estimated to ship onApril 02, 2025


Request a Bulk Order

vapor density

4.84 (−7 °C, vs air)

Quality Level

vapor pressure

4 mmHg ( −6.7 °C)

form

liquid

optical activity

[α]20/D −40 to +48°

quality

purified

impurities

≤0.0002% heavy metals (as Pb)
≤1.0% non-volatile matter

refractive index

n20/D 1.4670-1.4780

bp

153-175 °C (lit.)

mp

−55 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

InChI key

DCXXKSXLKWAZNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oil of turpentine is a mixture of volatile terpenic distillates of resin obtained from pines. It is generally composed of α-pinene, β-pinene, limonene and camphene. It possess antimicrobial[1] and larvicidal properties[2] and induces inflammation in high dosage.[3][4][5]

Application

Oil of turpentine is primarily used as a solvent for industrial applications, such as thinner for paints and varnishes.[6] Preparation of carbon nano-spheres (CNSs) by decomposition of turpentine oil has been reported.[7]

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Solvents.
Stoye D
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Larvicidal effect of Eucalyptus grandis essential oil and turpentine and their major components on Aedes aegypti larvae.
Lucia A, et al.
Journal of the American Mosquito Control Association, 23(3), 99-303 (2007)
Effects of Melilotus officinalis on acute inflammation.
Plesca?Manea L, et al.
Phytotherapy Research, 16(4), 316-319 (2002)
Evaluation of anti-inflammatory activity of alcoholic extract from Viola tricolor.
Toiu A, et al.
Revista medico-chirurgicala? a Societa?t?ii de Medici s?i Naturalis?ti Din Ias?i, 111(2), 525-529 (2007)
Synthesis of Turpentine Oil Based Carbon Nanospheres by Nebulized Spray Pyrolysis Method.
Arie AA, et al.
Journal of Nanoscience and Nanotechnology, 16(8), 8701-8704 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service