Skip to Content
Merck
All Photos(1)

Key Documents

SMB00279

Sigma-Aldrich

Chitosan dimer dihydrochloride

≥85% (HPLC), powder

Synonym(s):

Chitosan dimer dihydrochloride, Chitobiose dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$320.00

$320.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$320.00

About This Item

Empirical Formula (Hill Notation):
C12H24N2O9 · 2HCl
CAS Number:
Molecular Weight:
413.25
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

$320.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥85% (HPLC)

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](N)[C@H](O[C@@H]2[C@@H](CO)OC(O)[C@H](N)[C@H]2O)O[C@H](CO)[C@H]1O.Cl.Cl

InChI

1S/C12H24N2O9.2ClH/c13-5-9(19)10(4(2-16)21-11(5)20)23-12-6(14)8(18)7(17)3(1-15)22-12;;/h3-12,15-20H,1-2,13-14H2;2*1H/t3-,4-,5-,6-,7-,8-,9-,10-,11?,12+;;/m1../s1

InChI key

QBSWPIQMUDKRHR-SORUAGQPSA-N

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service