Skip to Content
Merck
All Photos(1)

Key Documents

912700

Sigma-Aldrich

Trimethyl chitosan

low molecular weight, degree of quaternization >50%

Synonym(s):

N,N,N-Trimethyl chitosan, Chitosan trimethyl, Mucoadhesive polymers, TMC Chitosan

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$552.00

$552.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$552.00

About This Item

Linear Formula:
(C9H18ClNO4)n
CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.23

$552.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

powder

color

light yellow to light brown

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Chitosan offers remarkable biological properties, it has been widely used in drug delivery and tissue engineering applications. Chitosan has good mucoadhesive properties due to its positive charge, which increases the adhesion to mucosa and facilitates drug penetration. Also, chitosan possesses hemostatic properties, which makes it a good candidate for wound dressing applications.
The quaternization of the primary amine increases the water solubility of chitosan and keeps chitosan soluble over a wide pH range. Among all the quaternized chitosans, N, N, N-trimethyl chitosan chloride (TMC) is the most widely applied in biomedical applications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and Characterization of Particles from Chitosan with Different Molecular Weights and Their Trimethyl Chitosan Derivatives for Nasal Immunization.
Boonyo W, et al.
The Journal of The Minerals, Metals & Materials Society, 18(2), 59-65 (2008)
Preparation and NMR characterization of highly substituted IV-trimethyl chitosan chloride.
Sieval A B, et al.
Carbohydrate Polymers, 36, 157-165 (1998)
Chitosan and Chitosan Derivatives in Drug Delivery and Tissue Engineering.
Riva R, et al.
Advances in Polymer Science, 244, 19-44 (2011)
J H Hamman et al.
Drug development and industrial pharmacy, 29(2), 161-172 (2003-03-22)
N-trimethyl chitosan chloride (TMC) is a polycation that enhances drug transport across epithelia by opening tight junctions. The degree of quaternization of TMC determines the number of positive charges available on the molecule for interactions with the negatively charged sites
Amir K Varkouhi et al.
Bioconjugate chemistry, 21(12), 2339-2346 (2010-11-06)
N,N,N-Trimethylated chitosan (TMC) is a biodegradable polymer emerging as a promising nonviral vector for nucleic acid and protein delivery. In the present study, we investigated whether the introduction of thiol groups in TMC enhances the extracellular stability of the complexes

Articles

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Microfluidic assembly can efficiently overcome the general disadvantages of polyamine nanoencapsulation of nucleic acids, such as a less defined morphology and composition, polydispersity, and poor reproducibility.

Questions

  1. Hi! We ordered and received this product, but we were inquiring to see if we could also get an exact molecular weight of this product as well. Thank you!

    1 answer
    1. The molecular weight is not determined, however the specification for Brookfield viscosity is 8 - 14 cP at 5% in water, 25°C. The term “low molecular weight ” refers to the raw material polymer used to manufacture this material.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service