Skip to Content
Merck
All Photos(1)

Key Documents

SMB00178

Sigma-Aldrich

Rhododendrol

≥95% (LC/MS-ELSD)

Synonym(s):

(R)-(-)-Rhododendrol, 4-Hydroxy-α-methyl-benzenepropanol, Betuligenol, Frambinol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$608.00

$608.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$608.00

About This Item

Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$608.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC(O)CCc1ccc(O)cc1

InChI

1S/C10H14O2/c1-8(11)2-3-9-4-6-10(12)7-5-9/h4-8,11-12H,2-3H2,1H3

InChI key

SFUCGABQOMYVJW-UHFFFAOYSA-N

General description

Natural product derived from plant source.

Biochem/physiol Actions

Rhododendrol is a natural phenolic compound that has been reported to prevent high-fat diet-induced elevation in body weight and to increase lipolysis in white adipocytes in male mice. RK has shown potential to increase dermal IGF-I production through sensory neuron activation, promoting hair growth and increasing skin elasticity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masatoshi Kondo et al.
Pigment cell & melanoma research, 29(5), 541-549 (2016-05-26)
Rhododendrol is a phenolic compound that shows a tyrosinase-dependent toxicity for melanocytes and occasionally induces a vitiligo-like skin depigmentation. The post-tyrosinase mechanisms determining melanocyte death or survival, however, are far from clear. Here, we find that rhododendrol treatment leads to
Rie Takagi et al.
Dermatology (Basel, Switzerland), 232(1), 44-49 (2015-11-28)
Rhododendrol, a phenolic compound contained in lightening/whitening cosmetics, can bind and inhibit tyrosinase and was reported to induce leukoderma in Japan. Only 2% of the cosmetics users are affected, and tacrolimus is effective in treatment of the condition. To test
Yu Gabe et al.
Journal of dermatological science, 91(3), 311-316 (2018-07-15)
Rhododendrol (4-(4-hydroxyphenyl)-2-butanol) has been used as a lightening/whitening cosmetic but was recently reported to induce leukoderma. Although rhododendrol has been shown to be transformed by tyrosinase to hydroxyl-rhododendrol, which is cytotoxic to melanocytes, its detailed mechanism of action including the
Minoru Sasaki et al.
Pigment cell & melanoma research, 27(5), 754-763 (2014-06-04)
Rhododendrol, an inhibitor of melanin synthesis developed for lightening/whitening cosmetics, was recently reported to induce a depigmentary disorder principally at the sites of repeated chemical contact. Rhododendrol competitively inhibited mushroom tyrosinase and served as a good substrate, while it also

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service