Skip to Content
Merck
All Photos(1)

Key Documents

SMB00174

Sigma-Aldrich

Cirsimaritin

≥90% (LC/MS-ELSD)

Synonym(s):

4′,5-Dihydroxy-6,7-dimethoxyflavone, Skrofulein

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$626.00

$626.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$626.00

About This Item

Empirical Formula (Hill Notation):
C17H14O6
CAS Number:
Molecular Weight:
314.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

$626.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥90% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1cc2OC(=CC(=O)c2c(O)c1OC)c3ccc(O)cc3

InChI

1S/C17H14O6/c1-21-14-8-13-15(16(20)17(14)22-2)11(19)7-12(23-13)9-3-5-10(18)6-4-9/h3-8,18,20H,1-2H3

InChI key

ZIIAJIWLQUVGHB-UHFFFAOYSA-N

General description

Natural product derived from plant source.

Biochem/physiol Actions

Cirsimaritin has been shown to inhibit tumor cell growth and induce mitochondrial apoptosis in GBC-SD cells. It was proposed that cirsimaritin-induced reactive oxygen species is the trigger for ER stress and down-regulate the phosphorylation of Akt.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Mohamed Ben Sghaier et al.
Environmental toxicology and pharmacology, 32(3), 336-348 (2011-10-19)
Fractionation of the chloroformic extracts from Teucrium ramosissimum leaves resulted in the isolation of three flavonoids: genkwanin (1), cirsimaritin (2) and 4',7-dimethoxy apigenin (4) and one sesquiterpene: β-eudesmol (3). The structures were determined using data obtained from (1)H and (13)C
J A Hasrat et al.
The Journal of pharmacy and pharmacology, 49(11), 1150-1156 (1997-12-24)
In traditional medicine Microtea debilis is used against proteinuria. In ligand-binding studies extracts of Microtea debilis have been shown to inhibit the binding of [3H]1,3-dipropyl-8-cyclopentylxanthine ([3H]DPCPX) to adenosine-A1 receptors in rat forebrain membranes. Subsequently, cirsimarin, a flavonoid, was isolated as
Jih-Pyang Wang et al.
Naunyn-Schmiedeberg's archives of pharmacology, 366(4), 307-314 (2002-09-19)
In this study, the cellular localization of the inhibitory effect of a natural flavonoid cirsimaritin against formyl-methionyl-leucyl-phenylalanine (fMLP)-induced respiratory burst in rat neutrophils was investigated. Cirsimaritin concentration-dependently inhibited the superoxide anion (O(*-)(2))generation and O(2) consumption (IC(50) 11.5+/-2.2 micro M and
Zhiwei Quan et al.
Cancer letters, 295(2), 252-259 (2010-04-03)
In this study, the anticancer effect of cirsimaritin, a natural flavonoid, against human gallbladder carcinoma cell line GBC-SD and the underlying mechanisms were investigated. Cirsimaritin inhibited the growth of tumor cells and induced mitochondrial apoptosis in GBC-SD cells. In addition
Effects of cirsimaritin, a flavone isolated from Artemisia judaica, on isolated guinea-pig ileum.
S S Abdalla et al.
Planta medica, 53(4), 322-324 (1987-08-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service