Skip to Content
Merck
All Photos(1)

Key Documents

SMB00129

Sigma-Aldrich

(15:3)-Anacardic acid

≥85% (LC/MS-ELSD)

Synonym(s):

Anacardic acid C15:3

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$423.00

$423.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
$423.00

About This Item

Empirical Formula (Hill Notation):
C22H30O3
CAS Number:
Molecular Weight:
342.47
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$423.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥85% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC(=O)c1c(O)cccc1CCCCCCC\C=C/C\C=C/CC=C

InChI

1S/C22H30O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-16-19-17-15-18-20(23)21(19)22(24)25/h2,4-5,7-8,15,17-18,23H,1,3,6,9-14,16H2,(H,24,25)/b5-4-,8-7-

InChI key

QUVGEKPNSCFQIR-UTOQUPLUSA-N

General description

Natural product derived from plant source.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service